    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KPNA4 Antibodies
          Proteintech
          KPNA4 Recombinant Rabbit Monoclonal Antibody (230098H1)
          View all (16) KPNA4 antibodies
          Tech Support
          Cite KPNA4 Recombinant Rabbit Monoclonal Antibody (230098H1)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          KPNA4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KPNA4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          KPNA4 Antibody (82898-2-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using KPNA4 antibody (82898-2-RR, Clone: 230098H1) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KPNA4 Antibody in Immunocytochemistry (ICC/IF)
          KPNA4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KPNA4 Antibody in Western Blot (WB)
          KPNA4 Antibody in Flow Cytometry (Flow)
          KPNA4 Antibody in Surface plasmon resonance (SPR)
          KPNA4 Recombinant Rabbit Monoclonal Antibody (230098H1)
          Product Details
          82898-2-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230098H1
          Immunogen
          KPNA4 fusion protein Ag3133
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MADNEKLDNQ RLKNFKNKGR DLETMRRQRN EVVVELRKNK RDEHLLKRRN VPHEDICEDS DIDGDYRVQN TSLEAIVQNA SSDNQGIQLS AVQAARKLLS SDRNPPIDDL IKSGILPILV HCLERDDNPS LQFEAAWALT NIASGTSEQT QAVVQSNAVP LFLRLLHSPH QNVCEQAVWA LGNIIGDGPQ CRDYVISLGV VKPLLSFISP SIPITFLRNV TWVMVNLCRH KDPPPPMETI QEILPALCVL IHHTDVNILV DTVWALSYLT DAGNEQIQMV IDSGIVPHLV PLLSHQEVKV QTAALRAVGN IVTGTDEQTQ VVLNCDALSH FPALLTHPKE KINKEAVWFL
          Target Information
          The nuclear import of karyophilic proteins is directed by short amino acid sequences termed nuclear localization signals (NLSs). Karyopherins, or importins, are cytoplasmic proteins that recognize NLSs and dock NLS-containing proteins to the nuclear pore complex. The protein encoded by this gene shares the sequence similarity with Xenopus importin-alpha and Saccharomyces cerevisiae Srp1. This protein is found to interact with the NLSs of DNA helicase Q1 and SV40 T antigen.
          Bioinformatics
          Protein Aliases: Importin alpha Q1; Importin subunit alpha-3; importin subunit alpha-4; importin-alpha-Q1; isoform 3; karyopherin (importin) alpha 4; karyopherin alpha 4; karyopherin alpha 4 (importin alpha 3); Karyopherin subunit alpha-4; MGC12217; MGC26703; predicted role in nuclear import; Qip1; Qip1 protein; unnamed protein product
          View more View less
          Gene Aliases: 1110058D08Rik; importin; IPOA3; KPNA4; QIP1; SRP3
          View more View less
          UniProt ID: (Human) O00629, (Mouse) O35343
          View more View less
          Entrez Gene ID: (Human) 3840, (Mouse) 16649, (Rat) 361959
          View more View less
          Function(s)
          protein binding nuclear localization sequence binding nuclear import signal receptor activity protein transporter activity transporter
          Process(es)
          protein import into nucleus NLS-bearing protein import into nucleus gene expression dopamine secretion protein transport positive regulation of hippo signaling response to hydrogen peroxide transport
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline