    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ZC3H11A Antibodies
          Proteintech
          ZC3H11A Recombinant Rabbit Monoclonal Antibody (230191D2)
          View all (13) ZC3H11A antibodies
          Tech Support
          Cite ZC3H11A Recombinant Rabbit Monoclonal Antibody (230191D2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          ZC3H11A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ZC3H11A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ZC3H11A Antibody (82912-5-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using ZC3H11A antibody (82912-5-RR, Clone: 230191D2) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZC3H11A Antibody in Immunocytochemistry (ICC/IF)
          ZC3H11A Antibody in Surface plasmon resonance (SPR)
          ZC3H11A Recombinant Rabbit Monoclonal Antibody (230191D2)
          Product Details
          82912-5-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230191D2
          Immunogen
          ZC3H11A fusion protein Ag23424
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: KSMQEVHIKT LEEIKLEKAL RVQQSSESST SSPSQHEATP GARRLLRITK RTGMKEEKNL QEGNEVDSQS SIRTEAKEAS GETTGVDITK IQVKRCETMR EKHMQKQQER EKSVLTPLRG DVASCNTQVA EKPVLTAVPG ITRHLTKRLP TKSSQKVEVE TSGIGDSLLN VKCAAQTLEK RGKAKPKVNV KPSVVKVVSS PKLAPKRKAV EMHAAVIAAV KPLSSSSVLQ EPPAKKAAVA VVPLVSEDKS VTVPEAENPR DSLVLPPTQS SSDSSPPEVS GPSSSQMSMK TRRLSSASTG KPPLSVEDDF EKLIWEISGG KLEAEIDLDP GKDEDDLLLE LSEMIDS
          Target Information
          Involved in nuclear mRNA export; probably mediated by association with the TREX complex. [UniProt]
          Bioinformatics
          Protein Aliases: unnamed protein product; Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH-type domain containing 11A
          View more View less
          Gene Aliases: KIAA0663; ZC3H11A; ZC3HDC11A
          View more View less
          UniProt ID: (Human) O75152
          View more View less
          Entrez Gene ID: (Human) 9877
          View more View less
          Function(s)
          RNA binding protein binding zinc ion binding metal ion binding
          Process(es)
          poly(A)+ mRNA export from nucleus mRNA transport
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline