    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MYOF Antibodies
          Proteintech
          MYOF Recombinant Rabbit Monoclonal Antibody (230213B6)
          View all (11) MYOF antibodies
          Tech Support
          Cite MYOF Recombinant Rabbit Monoclonal Antibody (230213B6)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MYOF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MYOF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MYOF Antibody (82917-7-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using MYOF antibody (82917-7-RR, Clone: 230213B6) at dilution of 1:250 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MYOF Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          82917-7-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:125-1:500
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230213B6
          Immunogen
          Peptide
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Target Information
          Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined.
          Bioinformatics
          Protein Aliases: fer-1-like 3, myoferlin; fer-1-like family member 3; Fer-1-like protein 3; ferlin; ferlin family member; Myoferlin; unnamed protein product
          View more View less
          Gene Aliases: FER1L3; HAE7; KIAA1207; MYOF
          View more View less
          UniProt ID: (Human) Q9NZM1
          View more View less
          Entrez Gene ID: (Human) 26509
          View more View less
          Function(s)
          calcium ion binding protein binding phospholipid binding metal ion binding
          Process(es)
          plasma membrane repair muscle contraction blood circulation regulation of neurotransmitter secretion
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline