    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • CYP7B1 Antibodies
          Proteintech
          CYP7B1 Recombinant Rabbit Monoclonal Antibody (230160H5)
          View all (8) CYP7B1 antibodies
          Tech Support
          Cite CYP7B1 Recombinant Rabbit Monoclonal Antibody (230160H5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          CYP7B1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CYP7B1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          CYP7B1 Antibody (82927-1-RR) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CYP7B1 antibody (82927-1-RR, Clone: 230160H5) at dilution of 1:1,000 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CYP7B1 Antibody in Immunocytochemistry (ICC/IF)
          CYP7B1 Antibody in Immunocytochemistry (ICC/IF)
          CYP7B1 Antibody in Western Blot (WB)
          CYP7B1 Antibody in Flow Cytometry (Flow)
          CYP7B1 Recombinant Rabbit Monoclonal Antibody (230160H5)
          Product Details
          82927-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunocytochemistry (ICC/IF)
          1:500-1:2,000
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230160H5
          Immunogen
          CYP7B1 fusion protein Ag19374
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: PFQYQLVIKN HKQLSFRVFS NKLLEKAFSI SQLQKNHDMN DELHLCYQFL QGKSLDILLE SMMQNLKQVF EPQLLKTTSW DTAELYPFCS SIIFEITFTT IYGKVIVCDN NKFISELRDD FLKFDDKFAY LVSNIPIELL GNVKSIREKI IKCFSSEKLA KMQGWSEVFQ SRQDVLEKYY VHEDLEIGAH HLGFLWASVA NTIPTMFWAM YYLLRHPEAM AAVRDEIDRL LQSTGQKKGS GFPIHLTREQ LDSLICLESS IFEALRLSSY STTIRFVEED LTLSSETGDY CVRKGDLVAI FPPVLHGD
          Target Information
          This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway of extrahepatic tissues, which converts cholesterol to bile acids. This enzyme likely plays a minor role in total bile acid synthesis, but may also be involved in the development of atherosclerosis, neurosteroid metabolism and sex hormone synthesis.
          Bioinformatics
          Protein Aliases: 24-hydroxycholesterol 7-alpha-hydroxylase; 25-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; cytochrome p450; Cytochrome P450 7B1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; Oxysterol 7-alpha-hydroxylase
          View more View less
          Gene Aliases: CBAS3; CP7B; CYP7B1; SPG5A
          View more View less
          UniProt ID: (Human) O75881
          View more View less
          Entrez Gene ID: (Human) 9420
          View more View less
          Function(s)
          monooxygenase activity iron ion binding protein binding steroid hydroxylase activity oxysterol 7-alpha-hydroxylase activity oxidoreductase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen heme binding 24-hydroxycholesterol 7alpha-hydroxylase activity 25-hydroxycholesterol 7alpha-hydroxylase activity metal ion binding
          Process(es)
          lipid metabolic process steroid biosynthetic process bile acid biosynthetic process steroid metabolic process cholesterol metabolic process sterol metabolic process intracellular estrogen receptor signaling pathway negative regulation of intracellular estrogen receptor signaling pathway B cell chemotaxis cholesterol homeostasis epithelial cell proliferation positive regulation of epithelial cell proliferation prostate gland epithelium morphogenesis
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline