    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • SLC22A12 Antibodies
          Proteintech
          URAT1 Recombinant Rabbit Monoclonal Antibody (230204B3)
          View all (7) SLC22A12 antibodies
          Tech Support
          Cite URAT1 Recombinant Rabbit Monoclonal Antibody (230204B3)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          URAT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          URAT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          URAT1 Antibody (82964-1-RR) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with 82964-1-RR (URat1 antibody) at dilution of 1:10,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          URAT1 Antibody in Western Blot (WB)
          URAT1 Antibody in Flow Cytometry (Flow)
          URAT1 Recombinant Rabbit Monoclonal Antibody (230204B3)
          Product Details
          82964-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230204B3
          Immunogen
          URAT1 fusion protein Ag27805
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 750
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: SELLDLVGGL GRFQVLQTMA LMVSIMWLCT QSMLENFSAA VPSHRCWAPL LDNSTAQASI LGSLSPEALL AISIPPGPNQ RPHQCRRFRQ PQWQLLDPNA TATSWSEADT EPCVDGWVYD RSIFTSTIVA KWNLVCDSHA LKPMAQSIYL AGILVGAAAC GPASDRFGRR LVLTWSYLQM AVMGTAAAFA PAFPVYCLFR FLLAFAVAGV MMNTGTLLME WTAARARPLV MTLNSLGFSF GHGLTAAVAY GVRDWTLLQL VVSVPFFLCF LYSWWLAESA RWLLTTGRLD WGLQELWRVA AINGKGAVQD TLTPEVLLSA MREELSMGQP PASLGTLLRM PGLRFRTC
          Target Information
          SLC22A12 is required for efficient urate re-absorption in the kidney. SLC22A12 regulates blood urate levels. SLC22A12 mediates saturable urate uptake by facilitating the exchange of urate against organic anions.The protein encoded by this gene is a urate transporter and urate-anion exchanger which regulates the level of urate in the blood. This protein is an integral membrane protein primarily found in kidney. Two transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: Organic anion transporter 4-like protein; Renal-specific transporter; RST; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; Solute carrier family 22 member 12; unnamed protein product; URAT1; Urate anion exchanger 1; urate anion transporter 1; urate transporter 1; Urate:anion antiporter SLC22A12
          View more View less
          Gene Aliases: AI987855; hURAT1; OAT4L; OATL4; RST; SLC22A12; Slc22al2; UAT; UNQ6453/PRO34004; URAT1
          View more View less
          UniProt ID: (Human) Q96S37, (Rat) Q3ZAV1, (Mouse) Q8CFZ5
          View more View less
          Entrez Gene ID: (Human) 116085, (Rat) 365398, (Mouse) 20521
          View more View less
          Function(s)
          urate transmembrane transporter activity transmembrane transporter activity PDZ domain binding molecular_function protein binding secondary carrier transporter
          Process(es)
          ion transport response to xenobiotic stimulus organic anion transport urate transport cellular homeostasis cellular response to insulin stimulus urate metabolic process transmembrane transport urate salt excretion biological_process organic acid transport response to drug transport organic acid transmembrane transport
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline