    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • RALGAPA1 Antibodies
          Proteintech
          GARNL1 Recombinant Rabbit Monoclonal Antibody (230165F11)
          View all (7) RALGAPA1 antibodies
          Tech Support
          Cite GARNL1 Recombinant Rabbit Monoclonal Antibody (230165F11)
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          GARNL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GARNL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GARNL1 Antibody (82971-1-RR) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with 82971-1-RR (GARNL1 antibody) at dilution of 1:5,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GARNL1 Antibody in Western Blot (WB)
          GARNL1 Antibody in Flow Cytometry (Flow)
          GARNL1 Antibody in Surface plasmon resonance (SPR)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          GARNL1 Recombinant Rabbit Monoclonal Antibody (230165F11)
          Product Details
          82971-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230165F11
          Immunogen
          GARNL1 fusion protein Ag31518
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.8 mg/mL
          Amount
          80 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LDLSDLPLDK LSEQKQKKHK GKGVGHEFQK VSVDKSFSRG WSRDQPGQAP MRQRSATTTG SPGTEKARSI VRQKTVDIDD AQILPRSTRV RHFSQSEETG NEVFGALNEE QPLPRSSSTS DILEPFTVER AKVNKEDMSQ K
          Target Information
          RALGAPA1 is a catalytic subunit of the heterodimeric RalGAP1 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
          Bioinformatics
          Protein Aliases: GAP-related interacting protein to E12; GTPase activating Rap/RanGAP domain-like 1; Ral GTPase activating protein catalytic alpha subunit 1; Ral GTPase activating protein, alpha subunit 1 (catalytic); tuberin-like protein 1; unnamed protein product
          View more View less
          Gene Aliases: GARNL1; GRIPE; NEDHRIT; p240; RalGAPalpha1; TULIP1
          View more View less
          UniProt ID: (Human) Q6GYQ0
          View more View less
          Entrez Gene ID: (Human) 253959
          View more View less
          Function(s)
          GTPase activator activity protein binding protein heterodimerization activity
          Process(es)
          small GTPase mediated signal transduction regulation of small GTPase mediated signal transduction activation of GTPase activity
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Thermo Fisher Scientific India Pvt Ltd.
          Registered office address:
          402, 403, 404, Delphi 'B' Wing,
          Hiranandani Business Park,
          Powai, Mumbai – 400 076
          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100MH2000PTC126872
          Invitrogen BioServices (India) Pvt Ltd
          Registered office address:
          First Technology Place, No 3, EPIP,
          Whitefield, Bangalore – 560 079
          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100KA2004PTC035330
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline