    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • ARID1B Antibodies
          Proteintech
          ARID1B Recombinant Rabbit Monoclonal Antibody (230222A11)
          View all (17) ARID1B antibodies
          Tech Support
          Cite ARID1B Recombinant Rabbit Monoclonal Antibody (230222A11)
          • Antibody Testing Data (2)
          ARID1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ARID1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ARID1B Antibody (82979-6-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using ARID1B antibody (82979-6-RR, Clone: 230222A11) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1).CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ARID1B Antibody in Immunocytochemistry (ICC/IF)
          ARID1B Antibody in Surface plasmon resonance (SPR)
          ARID1B Recombinant Rabbit Monoclonal Antibody (230222A11)
          Product Details
          82979-6-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230222A11
          Immunogen
          ARID1B fusion protein Ag30643
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: PAKRHEGDMY NMQYSSQQQE MYNQYGGSYS GPDRRPIQGQ YPYPYSRERM QGPGQIQTHG IPPQMMGGPL QSSSSEGPQQ NMWAARNDMP YPYQNRQGPG GPTQAPPYPG MNRTDDMMVP DQRINHESQW PSHVSQRQP
          Target Information
          This locus encodes an AT-rich DNA interacting domain-containing protein. The encoded protein is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. The protein encoded by this locus is similar to AT-rich interactive domain-containing protein 1A. These two proteins function as alternative, mutually exclusive ARID-subunits of the SWI/SNF complex. The associated complexes play opposing roles. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq, Feb 2012].
          Bioinformatics
          Protein Aliases: ARID domain-containing protein 1B; AT rich interactive domain 1B (SWI1-like); AT-rich interactive domain-containing protein 1B; BAF250B; BRG1-associated factor 250b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; ELD (eyelid)/OSA protein; hOsa2; Osa homolog 2; p250R
          View more View less
          Gene Aliases: 6A3-5; ARID1B; BAF250B; BRIGHT; CSS1; DAN15; ELD/OSA1; KIAA1235; MRD12; OSA2; P250R
          View more View less
          UniProt ID: (Human) Q8NFD5
          View more View less
          Entrez Gene ID: (Human) 57492
          View more View less
          Function(s)
          DNA binding transcription coactivator activity protein binding actin or actin-binding cytoskeletal protein
          Process(es)
          transcription, DNA-templated nervous system development covalent chromatin modification chromatin-mediated maintenance of transcription
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0