  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • TMEM132E Antibodies
          Proteintech
          TMEM132E Recombinant Rabbit Monoclonal Antibody (230525E12)
          View all (3) TMEM132E antibodies
          Tech Support
          Cite TMEM132E Recombinant Rabbit Monoclonal Antibody (230525E12)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TMEM132E Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TMEM132E Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TMEM132E Antibody (82983-2-RR) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with 82983-2-RR (TMEM132E antibody) at dilution of 1:2,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TMEM132E Antibody in Western Blot (WB)
          TMEM132E Recombinant Rabbit Monoclonal Antibody (230525E12)
          Product Details
          82983-2-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230525E12
          Immunogen
          TMEM132E fusion protein Ag26772
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 750
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MILPDRRSVR ESEDEDEEEE ERRQSASRGC TLQYQHATLQ VFTQFHTTSS EGTDQVVTML GPDWLVEVTD LVSDFMRVGD PRVAHMVDSS TLAGLEPGTT PFKVVSPLTE AVLGETLLTV TEEKVSITQL QAQVVASLAL SLRPSPGSSH TILATTAAQQ TLSFLKQEAL LSLWLSYSDG TTAPLSLYSP RDYGLLVSSL DEHV
          Target Information
          Required for normal inner ear hair cell function and hearing.
          Bioinformatics
          Protein Aliases: Transmembrane protein 132E
          View more View less
          Gene Aliases: DFNB99; TMEM132E
          View more View less
          UniProt ID: (Human) Q6IEE7
          View more View less
          Entrez Gene ID: (Human) 124842
          View more View less
          Process(es)
          posterior lateral line neuromast hair cell development
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline