    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • CCDC23 Antibodies
          Proteintech
          CCDC23 Recombinant Rabbit Monoclonal Antibody (230175B2)
          View all (2) CCDC23 antibodies
          Tech Support
          Cite CCDC23 Recombinant Rabbit Monoclonal Antibody (230175B2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CCDC23 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CCDC23 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CCDC23 Antibody (82998-4-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using CCDC23 antibody (82998-4-RR, Clone: 230175B2) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1).CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CCDC23 Antibody in Immunocytochemistry (ICC/IF)
          CCDC23 Antibody in Immunocytochemistry (ICC/IF)
          CCDC23 Recombinant Rabbit Monoclonal Antibody (230175B2)
          Product Details
          82998-4-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230175B2
          Immunogen
          CCDC23 fusion protein Ag33650
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MDPPARKEKT KVKESVSRVE KAKQKSAQQE LKQRQRAEIY ALNRVMTELE QQQFDEFCKQ MQPPGE
          Bioinformatics
          Protein Aliases: Coiled coil domain-containing protein 23; coiled-coil domain containing 23; coiled-coil domain-containing protein 23; Small vasohibin-binding protein; unnamed protein product
          View more View less
          Gene Aliases: CCDC23; NEDAHM; SPG94; SVBP
          View more View less
          UniProt ID: (Human) Q8N300
          View more View less
          Entrez Gene ID: (Human) 374969
          View more View less
          Function(s)
          protein binding microtubule binding peptidase activator activity
          Process(es)
          proteolysis protein secretion negative regulation of endothelial cell migration negative regulation of protein ubiquitination axon development regulation of metallopeptidase activity
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline