    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • PCYT1A Antibodies
          Proteintech
          PCYT1A Recombinant Rabbit Monoclonal Antibody (230393B4)
          View all (17) PCYT1A antibodies
          Tech Support
          Cite PCYT1A Recombinant Rabbit Monoclonal Antibody (230393B4)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          PCYT1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PCYT1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          PCYT1A Antibody (83100-1-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed MCF-7 cells using PCYT1A antibody (83100-1-RR, Clone: 230393B4) at dilution of 1:400 and Multi-rAb CoraLite ® Plus 488-Goat Anti-Rabbit Recombinant Secondary Antibody (H+L) (RGAR002), CL594-phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PCYT1A Antibody in Immunocytochemistry (ICC/IF)
          PCYT1A Antibody in Western Blot (WB)
          PCYT1A Antibody in Western Blot (WB)
          PCYT1A Antibody in Flow Cytometry (Flow)
          PCYT1A Antibody in Surface plasmon resonance (SPR)
          PCYT1A Recombinant Rabbit Monoclonal Antibody (230393B4)
          Product Details
          83100-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230393B4
          Immunogen
          PCYT1A fusion protein Ag34268
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MDAQCSAKVN ARKRRKEAPG PNGATEEDGV PSKVQRCAVG LRQPAPFSDE IEVDFSKPYV RVTMEEASRG TPCERP
          Target Information
          This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
          Bioinformatics
          Protein Aliases: CCT A; CCT-alpha; Choline-phosphate cytidylyltransferase A; choline-phosphate cytidylyltransferase alpha; CT A; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; CTP:phosphocholine cytidylyltransferase-alpha; phosphate cytidylyltransferase 1, choline, alpha; Phosphorylcholine transferase A; phosphorylcholine transferase alpha; unnamed protein product
          View more View less
          Gene Aliases: CCTA; CCTalpha; CGL5; CT; CTA; CTalpha; CTPCT; Cttalpha; PCYT1; PCYT1A; SMDCRD
          View more View less
          UniProt ID: (Human) P49585, (Mouse) P49586
          View more View less
          Entrez Gene ID: (Human) 5130, (Mouse) 13026
          View more View less
          Function(s)
          catalytic activity choline-phosphate cytidylyltransferase activity protein binding calmodulin binding lipid binding transferase activity nucleotidyltransferase activity phosphatidylcholine binding identical protein binding protein homodimerization activity molecular function inhibitor activity transferase
          Process(es)
          lipid metabolic process phosphatidylcholine biosynthetic process CDP-choline pathway phospholipid biosynthetic process B cell proliferation isotype switching phosphatidylcholine metabolic process organophosphate biosynthetic process biosynthetic process
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline