    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FMO1 Antibodies
          Proteintech
          FMO1 Recombinant Rabbit Monoclonal Antibody (230455B11)
          View all (9) FMO1 antibodies
          Tech Support
          Cite FMO1 Recombinant Rabbit Monoclonal Antibody (230455B11)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          FMO1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FMO1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          FMO1 Antibody (83156-3-RR) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with 83156-3-RR (FMO1 antibody) at dilution of 1:10,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FMO1 Antibody in Western Blot (WB)
          FMO1 Recombinant Rabbit Monoclonal Antibody (230455B11)
          Product Details
          83156-3-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230455B11
          Immunogen
          FMO1 fusion protein Ag34267
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 750
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LLKHIQFKTK VCSVTKCSDS AVSGQWEVVT MHEEKQESAI FDAVMVCTGF LTNPYLPLDS FPGINAFKGQ YFHSRQYKHP DIFKDKRVLV IGMGNSGTDI AVEASHLAEK VFLSTTGGGW VISRIFDSGY PWDMVFMTRF QNMLRNSLPT PIVTWLMERK INNWLNHANY GLIPEDRTQL KEFVLNDELP GRIITGKVFI RPSIKEVKEN SVIFNNTSKE EPIDIIVFAT GYTFAFPFLD ESVVKVEDGQ ASLYKYIFPA HLQKPTLAII GLIKPLGSMI PTGETQARWA VRVLKGVNKL PPPSVMIEEI NARKENKPSW FGLCYCKALQ S
          Target Information
          Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics.
          Bioinformatics
          Protein Aliases: Dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1; flavin containing monooxygenase 1; Flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; Hepatic flavin-containing monooxygenase 1; Trimethylamine monooxygenase; unnamed protein product
          View more View less
          Gene Aliases: Fmo-1; FMO1; RFMO1A
          View more View less
          UniProt ID: (Human) Q01740, (Rat) P36365, (Mouse) P50285
          View more View less
          Entrez Gene ID: (Human) 2326, (Rat) 25256, (Mouse) 14261
          View more View less
          Function(s)
          monooxygenase activity N,N-dimethylaniline monooxygenase activity protein binding oxidoreductase activity trimethylamine monooxygenase activity hypotaurine monooxygenase activity flavin adenine dinucleotide binding NADP binding oxidoreductase oxygenase metabolite interconversion enzyme
          Process(es)
          sulfur amino acid catabolic process organic acid metabolic process xenobiotic metabolic process toxin metabolic process response to lipopolysaccharide taurine biosynthetic process negative regulation of fatty acid oxidation energy homeostasis response to osmotic stress drug metabolic process oxidation-reduction process NADPH oxidation
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline