    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • MURF3 Antibodies
          Proteintech
          TRIM54 Recombinant Rabbit Monoclonal Antibody (230391G5)
          View all (25) MURF3 antibodies
          Tech Support
          Cite TRIM54 Recombinant Rabbit Monoclonal Antibody (230391G5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          TRIM54 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRIM54 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          TRIM54 Antibody (83175-2-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed C2C12 cells using TRIM54 antibody (83175-2-RR, Clone: 230391G5) at dilution of 1:200 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRIM54 Antibody in Immunocytochemistry (ICC/IF)
          TRIM54 Antibody in Western Blot (WB)
          TRIM54 Antibody in Flow Cytometry (Flow)
          TRIM54 Antibody in Surface plasmon resonance (SPR)
          TRIM54 Recombinant Rabbit Monoclonal Antibody (230391G5)
          Product Details
          83175-2-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:14,000
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230391G5
          Immunogen
          TRIM54 fusion protein Ag15229
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.7 mg/mL
          Amount
          70 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: IAMLVAGNDR VQAVITQMEE VCQTIEDNSR RQKQLLNQRF ESLCAVLEER KGELLQALAR EQEEKLQRVR GLIRQYGDHL EASSKLVESA IQSMEEPQMA LYLQQAKELI NKVGAMSKVE LAGRPEPGYE SMEQFTVRVE HVAEMLRTID FQPGASGEEE EVAPDGEEGS AGPEEERPDG P
          Target Information
          The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
          Bioinformatics
          Protein Aliases: MuRF; MuRF-3; Muscle-specific RING finger protein; Muscle-specific RING finger protein 3; muscle-specific RING-finger protein; OTTHUMP00000122598; RING finger protein 30; Tripartite motif-containing protein 54; unnamed protein product
          View more View less
          Gene Aliases: 4930486E09Rik; 4930566I02Rik; MURF; MURF-3; MURF3; RNF30; TRIM54
          View more View less
          UniProt ID: (Human) Q9BYV2, (Mouse) Q9ERP3
          View more View less
          Entrez Gene ID: (Human) 57159, (Mouse) 58522
          View more View less
          Function(s)
          protein binding microtubule binding zinc ion binding metal ion binding ubiquitin protein ligase activity ubiquitin-protein ligase
          Process(es)
          microtubule-based process negative regulation of microtubule depolymerization signal transduction cell differentiation innate immune response multicellular organism development
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline