    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RPL29 Antibodies
          Proteintech
          RPL29 Recombinant Rabbit Monoclonal Antibody (240062F9)
          View all (14) RPL29 antibodies
          Tech Support
          Cite RPL29 Recombinant Rabbit Monoclonal Antibody (240062F9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          RPL29 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RPL29 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          RPL29 Antibody (83377-1-RR) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using RPL29 antibody (83377-1-RR, Clone: 240062F9) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RPL29 Antibody in Immunocytochemistry (ICC/IF)
          RPL29 Antibody in Immunocytochemistry (ICC/IF)
          RPL29 Antibody in Western Blot (WB)
          RPL29 Antibody in Flow Cytometry (Flow)
          RPL29 Antibody in Surface plasmon resonance (SPR)
          RPL29 Recombinant Rabbit Monoclonal Antibody (240062F9)
          Product Details
          83377-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240062F9
          Immunogen
          RPL29 fusion protein Ag34100
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAKSKNHTTH NQSRKWHRNG IKKPRSQRYE SLKGVDPKFL RNMRFAKKHN KKGLKKMQAN NAKAMSARAE AIKALVKPKE VKPKIPKGVS RKLDRLA
          Target Information
          Dual specificity protein phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the MAPK superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of this family of dual specificity phosphatases show distinct substrate specificities for MAPKs, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product binds to and inactivates p38 and SAPK/JNK, but not MAPK/ERK. Its subcellular localization is unique; it is evenly distributed in both the cytoplasm and the nucleus. This gene is widely expressed in various tissues and organs, and its expression is elevated by stress stimuli. Three transcript variants encoding two different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: 60S ribosomal protein L29; Cell surface heparin-binding protein HIP; heparin/heparan sulfate-binding protein; heparin/heparan sulfate-interacting protein; HP/HS-interacting protein; Large ribosomal subunit protein eL29; P23; ribosomal protein YL43 homologue; unnamed protein product
          View more View less
          Gene Aliases: eL29; HIP; HUMRPL29; L29; RPL29; RPL29_3_370; RPL29P10; Rpl43
          View more View less
          UniProt ID: (Human) P47914, (Rat) P25886, (Mouse) P47915
          View more View less
          Entrez Gene ID: (Human) 6159, (Rat) 29283, (Mouse) 19944
          View more View less
          Function(s)
          RNA binding structural constituent of ribosome heparin binding cadherin binding poly(A) RNA binding ribosomal protein translational protein
          Process(es)
          cytoplasmic translation translation embryo implantation cell proliferation cell-substrate adhesion multicellular organism growth
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline