    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • INTS4 Antibodies
          Proteintech
          INTS4 Recombinant Rabbit Monoclonal Antibody (240579H4)
          View all (7) INTS4 antibodies
          Tech Support
          Cite INTS4 Recombinant Rabbit Monoclonal Antibody (240579H4)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          INTS4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          INTS4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          INTS4 Antibody (83590-5-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using INTS4 antibody (83590-5-RR, Clone: 240579H4) at dilution of 1:250 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          INTS4 Antibody in Immunocytochemistry (ICC/IF)
          INTS4 Antibody in Immunocytochemistry (ICC/IF)
          INTS4 Antibody in Flow Cytometry (Flow)
          INTS4 Antibody in Surface plasmon resonance (SPR)
          INTS4 Recombinant Rabbit Monoclonal Antibody (240579H4)
          Product Details
          83590-5-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:125-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240579H4
          Immunogen
          INTS4 fusion protein Ag9116
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAAHLKKRVY EEFTKVVQPQ EEIATKKLRL TKPSKSAALH IDLCKATSPA DALQYLLQFA RKPVEAESVE GVVRILLEHY YKENDPSVRL KIASLLGLLS KTAGFSPDCI MDDAINILQN EKSHQVLAQL LDTLLAIGTK LPENQAIQMR LVDVACKHLT DTSHGVRNKC LQLLGNLGSL EKSVTKDAEG LAARDVQKII GDYFSDQDPR VRTAAIKAML QLHERGLKLH QTIYNQACKL LSDDYEQVRS AAVQLIWVVS QLYPESIVPI PSSNEEIRLV DDAFGKICHM VSDGSWVVRV QAAKLLGSME QVSSHFLEQT LDKKLMSDLR R
          Target Information
          INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.
          Bioinformatics
          Protein Aliases: Int4; Integrator complex subunit 4; MSTP093; unnamed protein product
          View more View less
          Gene Aliases: INT4; INTS4; MST093; MSTP093
          View more View less
          UniProt ID: (Human) Q96HW7
          View more View less
          Entrez Gene ID: (Human) 92105
          View more View less
          Function(s)
          protein binding protein binding, bridging
          Process(es)
          snRNA processing regulation of transcription elongation from RNA polymerase II promoter RNA polymerase II transcription initiation surveillance
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline