    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MTMR9 Antibodies
          Proteintech
          MTMR9 Recombinant Rabbit Monoclonal Antibody (240167G7)
          View all (11) MTMR9 antibodies
          Tech Support
          Cite MTMR9 Recombinant Rabbit Monoclonal Antibody (240167G7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          MTMR9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MTMR9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          MTMR9 Antibody (83694-4-RR) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using MTMR9 antibody (83694-4-RR, Clone: 240167G7) at dilution of 1:200 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MTMR9 Antibody in Immunocytochemistry (ICC/IF)
          MTMR9 Antibody in Western Blot (WB)
          MTMR9 Antibody in Flow Cytometry (Flow)
          MTMR9 Antibody in Flow Cytometry (Flow)
          MTMR9 Antibody in Surface plasmon resonance (SPR)
          Product Details
          83694-4-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240167G7
          Immunogen
          MTMR9 fusion protein Ag34134
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MLFEHAYAS QFGTFLGNNE SERCKLKLQQ KTMSLWSWVN QPSELSKFTN PLFEANNLVI WPSVAPQSLP LWEGIFLRWN RSSKYLDEAY EEMVNIIEYN KELQAKVNIL RRQLAELETE DGMQESP
          Target Information
          This gene encodes a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate.
          Bioinformatics
          Protein Aliases: DKFZP434K171; Inactive phosphatidylinositol 3-phosphatase 9; MGC126672; myotubularin related protein 8; Myotubularin-related protein 9; unnamed protein product
          View more View less
          Gene Aliases: C8orf9; LIP-STYX; MTMR8; MTMR9
          View more View less
          UniProt ID: (Human) Q96QG7
          View more View less
          Entrez Gene ID: (Human) 66036
          View more View less
          Function(s)
          protein binding protein phosphatase binding enzyme regulator activity
          Process(es)
          endocytosis negative regulation of autophagy positive regulation of phosphatase activity phosphatidylinositol dephosphorylation protein stabilization regulation of phosphatidylinositol dephosphorylation
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline