  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • hnRNP A2B1 Antibodies
          Proteintech
          HNRNPA2B1 Recombinant Rabbit Monoclonal Antibody (240831E3)
          View all (15) hnRNP A2B1 antibodies
          Tech Support
          Cite HNRNPA2B1 Recombinant Rabbit Monoclonal Antibody (240831E3)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          HNRNPA2B1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HNRNPA2B1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          HNRNPA2B1 Antibody (83773-7-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using HNRNPA2B1 antibody (83773-7-RR, Clone: 240831E3) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HNRNPA2B1 Antibody in Immunocytochemistry (ICC/IF)
          HNRNPA2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPA2B1 Antibody in Western Blot (WB)
          HNRNPA2B1 Antibody in Flow Cytometry (Flow)
          HNRNPA2B1 Antibody in Surface plasmon resonance (SPR)
          HNRNPA2B1 Recombinant Rabbit Monoclonal Antibody (240831E3)
          Product Details
          83773-7-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:800
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240831E3
          Immunogen
          HNRNPA2B1 fusion protein Ag6563
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEREKEQFRK LFIGGLSFET TEESLRNYYE QWGKLTDCVV MRDPASKRSR GFGFVTFSSM AEVDAAMAAR PHSIDGRVVE PKRAVAREES GKPGAHVTVK KLFVGGIKED TEEHHLRDYF EEYGKIDTIE IITDRQSGKK RGFGFVTFDD HDPVDKIVLQ KYHTINGHNA EVRKALSRQE MQEDLEVAIL EVAPVMEEEE EDMVVEDLDM ATRVGATEVV MTTMEEEIME VEITMILEII TSNLLTTVQ
          Target Information
          This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq, Jul 2008]
          Bioinformatics
          Protein Aliases: DKFZp779B0244; epididymis secretory sperm binding protein; FLJ22720; heterogeneous nuclear ribonucleoprotein A2; heterogeneous nuclear ribonucleoprotein B0a; heterogeneous nuclear ribonucleoprotein B0b; heterogeneous nuclear ribonucleoprotein B1; Heterogeneous nuclear ribonucleoproteins A2/B1; heterogeneous nuclear ribonucleoproteins A2/B1 {ECO:0000312|EMBL:BAF79676.1}; heterogenous nuclear ribonucleoprotein A2/B1; hnRNP A2 / hnRNP B1; hnRNP A2 protein; hnRNP A2/B1; hnRNP B1 protein; hnrnpa2b1 {ECO:0000250|UniProtKB:P22626}; HNRNPA2B1/MYC fusion; nuclear ribonucleoprotein particle A2 protein; OTTHUMP00000202465; OTTHUMP00000202466; unnamed protein product
          View more View less
          Gene Aliases: 9130414A06Rik; Hnrnp; hnrnp-A; HNRNPA2; HNRNPA2B1; HNRNPB1; HNRPA2; HNRPA2B1; HNRPB1; IBMPFD2; OPMD2; RNPA2; SNRPB1
          View more View less
          UniProt ID: (Human) P22626, (Rat) A7VJC3, (Mouse) O88569
          View more View less
          Entrez Gene ID: (Human) 3181, (Rat) 362361, (Mouse) 53379
          View more View less
          Function(s)
          nucleic acid binding RNA binding mRNA 3'-UTR binding protein binding miRNA binding identical protein binding single-stranded telomeric DNA binding DNA polymerase binding G-rich strand telomeric DNA binding molecular condensate scaffold activity N6-methyladenosine-containing RNA binding nucleotide binding regulatory region RNA binding poly(A) RNA binding pre-mRNA intronic binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter mRNA processing RNA splicing male gonad development lung development negative regulation of mRNA splicing, via spliceosome oligodendrocyte differentiation RNA transport response to mineralocorticoid mRNA splicing, via spliceosome mRNA export from nucleus primary miRNA processing DNA geometric change mRNA transport regulation of macromolecule metabolic process positive regulation of telomere maintenance via telomere lengthening miRNA transport
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline