    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TMT Antibodies
          Invitrogen
          TMT Monoclonal Antibody (25D5)
          Advanced Verification
          1 Published Figure
          9 References
          Tech Support
          Cite TMT Monoclonal Antibody (25D5)
          Additional Information:
          {{banner.description}}
          • Published Figures (1)
          • Advanced Verification (1)
          TMT Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TMT Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          TMT Antibody (90075) in WB
          The Biochemical journal 2017 - Published figure using TMT monoclonal antibody (Product # 90075) in Western Blot {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TMT Antibody in Western Blot (WB)
          TMT Antibody
          Product Details
          90075
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          View 8 publications 8 publications
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Immunoelectrophoresis (IE)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Chemical
          Published species
          Chemical, Tag
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          25D5
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          10 µg
          Purification
          Protein G
          Storage buffer
          PBS with 4% BSA
          Contains
          Kathon
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_10854708
          Product Specific Information
          Product # 90075 binds to immobilized TMT surface.
          Product # 90075 has been succesfully used in immunoprecipitation and Western blot applications.
          Target Information
          Tandem Mass Tag (TMT) reagents enable concurrent identification and multiplexed quantitation of proteins in different samples using tandem mass spectrometry (MS). Isobaric TMT reagents share identical strucures, including a peptide-reactive labeling group, a spacer arm and a MS/MS reporter. During the MS/MS analysis, each isobaric tag produces a unique reporter ion that makes quantitation possible. Amine-reactive NHS-ester isotopic duplex or six-plex isobaric TMT tags covalently react with amino termini and lysine residues of peptides and proteins. Cysteine-reactive Tandem Mass Tag (cysTMT) reagents react with reduced thiols of cysteine-containing peptides.
          Bioinformatics
          Protein Aliases: Tandem Mass Tag
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline