    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CHD4 Antibodies
          Active Motif
          Mi-2 beta (CHD4) Recombinant Mouse Monoclonal Antibody, AbFlex®
          View all (25) CHD4 antibodies
          Tech Support
          Cite Mi-2 beta (CHD4) Recombinant Mouse Monoclonal Antibody, AbFlex®
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Mi-2 beta (CHD4) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Mi-2 beta (CHD4) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Mi-2 beta (CHD4) Antibody (91276) in WB
          AbFlex® Mi-2 beta (CHD4) antibody (rAb) tested by Western blot 20 µg of Jurkat cell nuclear extract was run on SDS-PAGE and probed with AbFlex Mi-2 beta (CHD4) antibody at 0.5 µg/ml. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mi-2 beta (CHD4) Antibody in Western Blot (WB)
          Mi-2 beta (CHD4) Recombinant Mouse Monoclonal Antibody, AbFlex®
          Product Details
          91276
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.5-2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Immunogen
          This antibody was raised against a synthetic peptide derived from human Mi-2 beta.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Protein A
          Storage buffer
          32mM sodium acetate/140mM HEPES, pH 7.5, with 70mM NaCl, 30% glycerol
          Contains
          0.035% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          The product of this gene belongs to the SNF2/RAD54 helicase family. It represents the main component of the nucleosome remodeling and deacetylase complex and plays an important role in epigenetic transcriptional repression. Patients with dermatomyositis develop antibodies against this protein.
          Bioinformatics
          Protein Aliases: ATP-dependent chromatin remodeler CHD4; ATP-dependent helicase CHD4; CHD-4; Chromo domain-containing protein 4; Mi-2 autoantigen 218 kDa protein; Mi2-beta; MI2B; unnamed protein product
          View more View less
          Gene Aliases: CHD-4; CHD4; Mi-2b; Mi2-BETA; SIHIWES
          View more View less
          UniProt ID: (Human) Q14839
          View more View less
          Entrez Gene ID: (Human) 1108
          View more View less
          Function(s)
          nucleotide binding transcription cofactor binding DNA binding chromatin binding transcription corepressor activity protein binding ATP binding zinc ion binding hydrolase activity ATPase activity nucleosomal DNA binding histone binding histone deacetylase binding metal ion binding RNA polymerase II sequence-specific DNA binding transcription factor binding DNA-binding transcription factor binding ATP-dependent chromatin remodeler activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter double-strand break repair via homologous recombination chromatin organization chromatin remodeling negative regulation of gene expression oxygen transport regulation of cell fate specification negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated regulation of synapse assembly terminal button organization regulation of stem cell differentiation
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline