    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CD33 Antibodies
          NeoBiotechnologies
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Monoclonal Antibody (SIGLEC3/3600)
          View all (117) CD33 antibodies
          Tech Support
          Cite CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Monoclonal Antibody (SIGLEC3/3600)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody (945-MSM10-P0) in IHC (P)
          Formalin-fixed, paraffin-embedded human urothelial carcinoma stained with CD33 Mouse Monoclonal Antibody (SIGLEC3/3600). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in Peptide array (ARRAY)
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Antibody in SDS-PAGE (SDS-PAGE)
          CD33/SIGLEC3 (Myeloid Cell Surface Antigen) Monoclonal Antibody (SIGLEC3/3600)
          Product Details
          945-MSM10-P0
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-dependent
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1-2 µg/mL
          -
          Peptide Array (Array)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2c, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          SIGLEC3/3600
          Immunogen
          Recombinant fragment (around aa51-224) of human CD33 protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          20 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.
          Bioinformatics
          Protein Aliases: CD33; CD33 antigen (gp67); CD33 molecule transcript; FLJ00391; gp67; lacks exon 2 but includes exon 7a; Myeloid cell surface antigen CD33; Sialic acid-binding Ig-like lectin 3; Siglec-3; unnamed protein product
          View more View less
          Gene Aliases: CD33; CD33rSiglec; p67; SIGLEC-3; SIGLEC3
          View more View less
          UniProt ID: (Human) P20138
          View more View less
          Entrez Gene ID: (Human) 945
          View more View less
          Function(s)
          protein binding protein tyrosine phosphatase activator activity protein phosphatase binding carbohydrate binding sialic acid binding signaling receptor activity
          Process(es)
          immune response-inhibiting signal transduction cell adhesion signal transduction cell-cell signaling negative regulation of cell proliferation negative regulation of interleukin-1 beta production negative regulation of interleukin-8 production negative regulation of tumor necrosis factor production Fc-gamma receptor signaling pathway positive regulation of protein secretion cell-cell adhesion negative regulation of monocyte activation
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline