    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • CD66b (CEACAM8) Antibodies
          Proteintech
          CEACAM8/CD66b Recombinant Rabbit Monoclonal Antibody (230353B7)
          View all (18) CD66b (CEACAM8) antibodies
          Tech Support
          Cite CEACAM8/CD66b Recombinant Rabbit Monoclonal Antibody (230353B7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CEACAM8/CD66b Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CEACAM8/CD66b Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CEACAM8/CD66b Antibody (98012-1-RR) in Flow
          1x10^6 human peripheral blood leukocytes were surface stained with 0.25 µg Anti-Human CEACAM5/8 (CD66b/e) Rabbit Recombinant Antibody (98012-1-RR, Clone: 230353B7) or 0.25 µg Isotype Control, and PE-conjugated Goat Anti-Rabbit IgG. Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CEACAM8/CD66b Antibody in Flow Cytometry (Flow)
          CEACAM8/CD66b Antibody in Surface plasmon resonance (SPR)
          Product Details
          98012-1-RR
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230353B7
          Immunogen
          Recombinant protein
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • CoraLite Plus 594
          • CoraLite Plus 647
          • CoraLite Plus 750
          • CoraLitePlus 405
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          Target Information
          This gene encodes a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregnancy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of all variants has not been defined.
          Bioinformatics
          Protein Aliases: Carcinoembryonic antigen CGM6; carcinoembryonic antigen gene family member 6; carcinoembryonic antigen related cell adhesion molecule 8; Carcinoembryonic antigen-related cell adhesion molecule 8; CD66b; CD67 antigen; CEA cell adhesion molecule 8; Cell adhesion molecule CEACAM8; Non-specific cross-reacting antigen NCA-95; unnamed protein product
          View more View less
          Gene Aliases: CD66b; CD67; CEACAM8; CGM6; NCA-95
          View more View less
          UniProt ID: (Human) P31997
          View more View less
          Entrez Gene ID: (Human) 1088
          View more View less
          Function(s)
          protein binding protein heterodimerization activity
          Process(es)
          immune response cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0