    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Fluorescein/Oregon Green Antibodies
          Invitrogen
          Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 488
          4 Published Figures
          37 References
          View all (5) Fluorescein/Oregon Green antibodies
          Tech Support
          Cite Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 488
          Additional Information:
          {{banner.description}}
          • Published Figures (4)
          Fluorescein/Oregon Green Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Fluorescein/Oregon Green Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Fluorescein/Oregon Green Antibody (A-11096) in IHC
          Psychoneuroendocrinology 2016 - Published figure using Fluorescein/Oregon Green polyclonal antibody (Product # A-11096) in Immunohistochemistry {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Fluorescein/Oregon Green Antibody in Immunohistochemistry (IHC)
          Fluorescein/Oregon Green Antibody in Immunohistochemistry (IHC)
          Fluorescein/Oregon Green Antibody in Immunohistochemistry (IHC)
          Fluorescein/Oregon Green Antibody in Immunohistochemistry (IHC)
          Product Details
          A-11096
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          -
          View 24 publications 24 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          -
          View 1 publication 1 publication
          Immunohistochemistry (Frozen) (IHC (F))
          -
          View 2 publications 2 publications
          Immunocytochemistry (ICC/IF)
          -
          View 4 publications 4 publications
          Flow Cytometry (Flow)
          -
          View 1 publication 1 publication
          ELISA (ELISA)
          Assay-dependent
          View 2 publications 2 publications
          Miscellaneous PubMed (Misc)
          -
          View 3 publications 3 publications
          Product Specifications
          Species Reactivity
          Chemical
          Published species
          Chemical, Mouse
          Host/Isotype
          Goat / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Fluorescein/Oregon Green
          Conjugate
          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          Excitation/Emission Max
          499/520 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          500 µg
          Purification
          purified
          Storage buffer
          0.1M potassium phosphate, pH 8
          Contains
          5mM sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_221558
          Product Specific Information
          We offer a variety of anti-fluorescent dye antibodies that recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria, or other biomolecules have been internalized by endocytic or pinocytic processes.
          In addition to their utility in quenching the fluorescence of extracellular fluorescein, the high affinity of Invitrogen's anti-fluorescein antibodies for both fluorescein and the structurally related Oregon GreenTM dye makes them ideal for immunochemical applications. Harmer and Samuel report the fluorescein-anti-fluorescein system provides a sensitive alternative to the biotin-streptavidin methods conventionally used for enzyme-linked immunosorbent assays (ELISAs). Fluorescein has also been found to be an excellent hapten for FISH, yielding sensitive results with extremely low background levels. The Alexa Fluor 488 conjugate can be used to amplify the fluorescence signal while still allowing fluorescein-compatible optics. The approximate absorption (Abs) and fluorescence emission (Em) maxima in nm: 495/519.
          Our anti-dye antibodies can be used in many different applications. It is a good practice to centrifuge the labeled antibody solutions briefly in a microcentrifuge before use; only the supernatant should then be added to the experiment. This step will eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
          Target Information
          Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
          Bioinformatics
          Protein Aliases: FITC
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline