    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TNFRSF14 (HVEM) Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          GenScript
          HVEM Monoclonal Antibody (10C6)
          View all (52) TNFRSF14 (HVEM) antibodies
          Tech Support
          Cite HVEM Monoclonal Antibody (10C6)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          HVEM Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          HVEM Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          HVEM Antibody (A01959-40) in Flow
          Flow cytometric analysis of peripheral blood mononuclear cell (PBMCs) binding with Human HVEM Antibody(10C6), mAb, Mouse (GenScript, A01959; blue curve) or binding with an isotype control antibody (red curve), or binding with PBS solution as a negative control (black curve). Antibody working concentration: 1 µg/mL, 2.5x105 cells/reaction. The signal was developed with R-PE conjugated anti-mouse IgG. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HVEM Antibody in Flow Cytometry (Flow)
          Product Details
          A01959-40
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          1- 5 µg/mL
          -
          ELISA (ELISA)
          0.01-0.05 µg/mL
          -
          Product Specifications
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          10C6
          Immunogen
          Recombinant HVEM protein
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Amount
          40 µg
          Storage conditions
          -20°C or -80°C if preferred
          Shipping conditions
          Wet ice
          Product Specific Information
          Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
          HVEM, also known as CD270, is a type 1 transmembrane protein. It is a member of the TNFR superfamily with a molecular weight of 30kDa. HVEM is expressed on hematopoietic cells such as T cells, B cells, dendritic cells, monocytes, neutrophils and NK cells. It participates in regulating lymphocyte activation and homeostasis in immune responses. HVEM activates NF-kB through the TNF-related cytokine LIGHT during T cell activation.
          Target Information
          TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline