    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PD-1 (CD279) Antibodies
          GenScript
          Nivolumab Recombinant Rabbit Monoclonal Antibody (40F11), Biotin, MonoRab™
          View all (283) PD-1 (CD279) antibodies
          Tech Support
          Cite Nivolumab Recombinant Rabbit Monoclonal Antibody (40F11), Biotin, MonoRab™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Nivolumab Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          Nivolumab Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Nivolumab Antibody (A01989-40) in ELISA
          Standard curve of Nivolumab Sandwich ELISA. The Nivolumab Sandwich ELISA assay is developed by using MonoRabAnti-Nivolumab Antibody (33H4), mAb, Rabbit (GenScript, A01987-40) and MonoRabAnti-Nivolumab Antibody (40F11)[Biotin] (Genscript, A01989-40) as the capture and detection antibodies, respectively. The sensitivity of detecting Nivolumab is up to 78 pg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Nivolumab Antibody in ELISA (ELISA)
          Nivolumab Antibody in ELISA (ELISA)
          Product Details
          A01989-40
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          Assay-dependent
          -
          Inhibition Assays (Inhibition)
          10-50 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          Mammalian cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          40F11
          Immunogen
          Nivolumab
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Lyophilized
          Amount
          40 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL.
          Nivolumab is a fully human monoclonal IgG4 antibody that is approved by the U.S. Food and Drug Administration for the treatment of bladder cancer. It binds to the programmed cell death protein 1 (PD-1) on the surface of activated T cells. Nivolumab functions as PD-1 inhibitor for targeted immunotherapy.GenScript MonoRab™ Anti-Nivolumab Antibody (40F11), mAb, Rabbit is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a rabbit immunized with Nivolumab.
          Upon reconstitution, it can be stored for 2-3 weeks at 2-8 °C or for up to 12 months at -20 °C or below.
          Target Information
          Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
          Bioinformatics
          Protein Aliases: CD279; CD279; PD-1; hPD1; PD-1; PD1; alternatively spliced; PDCD1; programmed cell death 1 protein; Programmed cell death protein 1; Protein PD-1; Protein PD1; sCD279; soluble CD279; systemic lupus erythematosus susceptibility 2; unnamed protein product
          View more View less
          Gene Aliases: AIMTBS; CD279; hPD-1; hPD-l; hSLE1; PD-1; PD1; PDCD1; SLEB2
          View more View less
          UniProt ID: (Human) Q15116
          View more View less
          Entrez Gene ID: (Human) 5133
          View more View less
          Function(s)
          protein binding signaling receptor activity
          Process(es)
          B cell apoptotic process adaptive immune response immune system process negative regulation of tolerance induction negative regulation of T cell mediated immune response to tumor cell negative regulation of B cell apoptotic process apoptotic process humoral immune response regulation of immune response negative regulation of immune response negative regulation of T cell activation negative regulation of multicellular organismal process positive regulation of T cell apoptotic process regulatory T cell apoptotic process
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline