    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • HA Tag Antibodies
          GenScript
          HA tag Recombinant Rabbit Monoclonal Antibody (109B2), Biotin, MonoRab™
          View all (98) HA Tag antibodies
          Tech Support
          Cite HA tag Recombinant Rabbit Monoclonal Antibody (109B2), Biotin, MonoRab™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          HA tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HA tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          HA tag Antibody (A02002-40) in WB
          Western blot analysis of MonoRab™ HA tag Antibody (109B2) [Biotin], mAb, Rabbit with HA-tag fusion proteins. Loading: Lane 1 : 25 ng N-terminal HA-tagged fusion protein Lane 2 : 10 ng N-terminal HA-tagged fusion protein Lane 3 : 5 ng N-terminal HA-tagged fusion protein Lane 4 : 1 ng N-terminal HA-tagged fusion protein Lane 5: 25 ng C-terminal HA-tagged fusion protein Lane 6: 10 ng C-terminal HA-tagged fusion protein Lane 7: 5 ng C-terminal HA-tagged fusion protein Lane 8: 1 ng C-terminal HA-tagged fusion protein Primary Antibody: MonoRabᵀᴹ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HA tag Antibody in Western Blot (WB)
          Product Details
          A02002-40
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.2-1 µg/mL
          -
          ELISA (ELISA)
          0.05-0.2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Rabbit / IgG
          Expression System
          Mammalian cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          109B2
          Immunogen
          A synthetic peptide containing the influenza hemagglutinin epitope (YPYDVPDYA). conjugated to KLH
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Lyophilized
          Amount
          40 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL.
          The rabbit immune system generates antibody diversity and optimizes affinity. GenScript utilizes MonoRab™ technology to generate high affinity and specific monoclonal rabbit antibodies. The HA tag is composed of several amino acids containing YPYDVPDYA and does not appear to interfere with the bioactivity or the biodistribution of recombinant proteins.GenScript MonoRab™ HA tag Antibody (109B2) [Biotin], mAb, Rabbit is the biotin conjugated form of MonoRab™ HA tag Antibody (109B2), mAb, Rabbit. This antibody is suitable for detecting the expression level of HA tag fusion proteins.
          Upon reconstitution, it can be stored for 2-3 weeks at 2-8 °C or for up to 12 months at -20 °C or below.
          Target Information
          HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
          Bioinformatics
          Protein Aliases: HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline