            • Produtos Customizados & Projetos
          • Secondary Antibodies ›
          • Goat anti-Mouse IgG2b Secondary Antibodies
          Invitrogen
          Goat anti-Mouse IgG2b Secondary Antibody, Alexa Fluor™ 633
          Advanced Verification
          View all (19) Goat anti-Mouse IgG2b antibodies
          Tech Support
          Cite Goat anti-Mouse IgG2b Secondary Antibody, Alexa Fluor™ 633
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          • Advanced Verification (1)
          Mouse IgG2b Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Mouse IgG2b Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Mouse IgG2b Secondary Antibody (A-21146) in ICC/IF
          Immunofluorescence analysis of Goat anti-Mouse IgG2b Secondary Antibody, Alexa Fluor™ 633 (Product # A-21146) was performed using LNCaP (positive model) and HeLa (negative model) cells stained with CD10 Monoclonal Antibody (Product # 14-0106-82). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, blocked with 1% BSA for 1 hour and labeled with... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse IgG2b Secondary Antibody in Immunocytochemistry (ICC/IF)
          Mouse IgG2b Secondary Antibody in Western Blot (WB)
          Mouse IgG2b Secondary Antibody in Western Blot (WB)
          Mouse IgG2b Secondary Antibody
          Product Details
          A-21146
          Applications
          Tested Dilution
          Western Blot (WB)
          1:10,000
          Immunohistochemistry (IHC)
          1-10 µg/mL
          Immunocytochemistry (ICC/IF)
          1-10 µg/mL
          Flow Cytometry (Flow)
          1-10 µg/mL
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Goat / IgG
          Class
          Polyclonal
          Type
          Secondary Antibody
          Immunogen
          Mouse IgG2b
          Conjugate
          Alexa Fluor™ 633 Alexa Fluor™ 633 Alexa Fluor™ 633
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 647 
          • Alexa Fluor Plus 680 
          • Alexa Fluor Plus 800 
          • PE 
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          631/650 nm View spectra spectra
          Form
          Liquid
          Concentration
          2 mg/mL
          Amount
          500 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.5
          Contains
          5mM sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2535782
          Target
          IgG2b
          Cross Adsorption
          Prior to conjugation, antibody was solid phase adsorbed using mouse IgG1, IgG2a and IgG3 immunosorbents to remove cross reactive antibody.
          Antibody Form
          Whole Antibody
          Product Specific Information
          Fluorescence of this long-wavelength Alexa Fluor dye is not visible by looking through a conventional fluorescence microscope.
          Product will be shipped at Room Temperature.
          Target Information
          Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody A-21146 on CiteAb
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-4kcjf:80/100.66.78.36:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline