    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • RPRD2 Antibodies
          Bethyl Laboratories
          KIAA0460 Polyclonal Antibody
          View all (6) RPRD2 antibodies
          Tech Support
          Cite KIAA0460 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          KIAA0460 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          KIAA0460 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          KIAA0460 Antibody (A300-031A-T) in WB
          Detection of human KIAA0460 by western blot. Samples: Whole cell lysate (50 µg) from HeLa and HEK293T cells prepared using NETN lysis buffer. Antibody: Affinity purified goat anti-KIAA0460 antibody (Product # A300-031A) (lot A300-031A-2) used for WB at 0.2 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KIAA0460 Antibody in Western Blot (WB)
          KIAA0460 Polyclonal Antibody
          Product Details
          A300-031A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Goat / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 700 and 750 of human KIAA0460.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          RPRD2 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; RNA polymerase II, holoenzyme; RNA polymerase II complex binding; mRNA 3'-end processing; snRNA transcription by RNA polymerase II.
          Bioinformatics
          Protein Aliases: Regulation of nuclear pre-mRNA domain-containing protein 2; unnamed protein product
          View more View less
          Gene Aliases: HSPC099; KIAA0460; RPRD2
          View more View less
          UniProt ID: (Human) Q5VT52
          View more View less
          Entrez Gene ID: (Human) 23248
          View more View less
          Function(s)
          RNA polymerase II core binding RNA polymerase II C-terminal domain binding
          Process(es)
          mRNA 3'-end processing
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline