    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • NCAPG2 Antibodies
          Bethyl Laboratories
          CAP-G2 Polyclonal Antibody
          View all (5) NCAPG2 antibodies
          Tech Support
          Cite CAP-G2 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CAP-G2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CAP-G2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CAP-G2 Antibody (A300-605A-T) in WB
          Detection of human CAP-G2 by western blot. Samples: Whole cell lysate (50 µg) from HeLa, HEK293T, MCF-7, K-562, and U2OS cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-CAP-G2 antibody (Product # A300-605A lot 4) used for WB at 0.1 µg/mL. Detection: Chemiluminescence with an exposure time of 10 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CAP-G2 Antibody in Western Blot (WB)
          CAP-G2 Antibody in Immunoprecipitation (IP)
          CAP-G2 Polyclonal Antibody
          Product Details
          A300-605A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1100 and the C-terminus (residue 1143) of human Chromosome-Associated Protein G2.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.20 mg/mL
          Amount
          2 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          TBS, pH 7.0 to 8.0, with 0.1% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          This gene encodes a protein that belongs to the Condensin2nSMC family of proteins. The encoded protein is a regulatory subunit of the condensin II complex which, along with the condensin I complex, plays a role in chromosome assembly and segregation during mitosis. A similar protein in mouse is required for early development of the embryo. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Aug 2013].
          Bioinformatics
          Protein Aliases: CAP-G2; Chromosome-associated protein G2; Condensin-2 complex subunit G2; Leucine zipper protein 5; more than blood homolog; Non-SMC condensin II complex subunit G2; unnamed protein product
          View more View less
          Gene Aliases: 3KS; CAP-G2; CAPG2; hCAP-G2; LUZP5; MTB; NCAPG2
          View more View less
          UniProt ID: (Human) Q86XI2
          View more View less
          Entrez Gene ID: (Human) 54892
          View more View less
          Function(s)
          protein binding bHLH transcription factor binding histone H4K20me1 reader activity
          Process(es)
          mitotic sister chromatid segregation inner cell mass cell proliferation chromatin organization transcription from RNA polymerase II promoter erythrocyte differentiation chromosome condensation cell division positive regulation of chromosome segregation positive regulation of chromosome separation positive regulation of chromosome condensation
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline