    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • DTX3L Antibodies
          Bethyl Laboratories
          DTX3L/BBAP Polyclonal Antibody
          View all (7) DTX3L antibodies
          Tech Support
          Cite DTX3L/BBAP Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          DTX3L/BBAP Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          DTX3L/BBAP Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          DTX3L/BBAP Antibody (A300-833A-T) in IP
          Detection of human DTX3L/BBAP by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HeLa cells. Antibodies: Affinity purified rabbit anti-DTX3L/BBAP antibody BL3622 (Product # A300-833A) used for IP at 3 µg/mg lysate. DTX3L/BBAP was also immunoprecipitated by rabbit anti-DTX3L/BBAP antibodies BL3623 and BL3624 (Product #A300-834A), which recognize downstream epitopes. For blotting immunoprecipitated DTX3L/BBAP, BL3624 was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DTX3L/BBAP Antibody in Immunoprecipitation (IP)
          DTX3L/BBAP Polyclonal Antibody
          Product Details
          A300-833A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-5 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 75 and 125 of human Deltex 3-Like Protein (B-Lymphoma- and BAL-Associated Protein).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          Component of the shelterin complex (telosome) that is involved in the regulation of telomere length and protection. Shelterin associates with arrays of double-stranded TTAGGG repeats added by telomerase and protects chromosome ends. Without its protective activity, telomeres are no longer hidden from the DNA damage surveillance and chromosome ends are inappropriately processed by DNA repair pathways. Promotes binding of POT1 to single-stranded telomeric DNA. Modulates the inhibitory effects of POT1 on telomere elongation. The ACD-POT1 heterodimer enhances telomere elongation by recruiting telomerase to telomeres and increasing its processivity. May play a role in organogenesis.
          Bioinformatics
          Protein Aliases: B-lymphoma- and BAL-associated protein; BBAP; deltex 3 like, E3 ubiquitin ligase; deltex 3-like; E3 ubiquitin-protein ligase DTX3L; Protein deltex-3-like; Rhysin-2; Rhysin2; RING-type E3 ubiquitin transferase DTX3L; unnamed protein product
          View more View less
          Gene Aliases: BBAP; DTX3L; RNF143
          View more View less
          UniProt ID: (Human) Q8TDB6
          View more View less
          Entrez Gene ID: (Human) 151636
          View more View less
          Function(s)
          ubiquitin-protein transferase activity enzyme inhibitor activity protein binding zinc ion binding transferase activity enzyme binding histone binding ubiquitin-like protein ligase binding metal ion binding ubiquitin protein ligase activity STAT family protein binding protein ADP-ribosyltransferase-substrate adaptor activity histone ubiquitin ligase activity histone H4K91 ubiquitin ligase activity
          Process(es)
          DNA damage checkpoint positive regulation of defense response to virus by host immune system process DNA repair double-strand break repair chromatin organization ubiquitin-dependent protein catabolic process cellular response to DNA damage stimulus Notch signaling pathway endosome to lysosome transport protein transport protein ubiquitination positive regulation of protein binding positive regulation of chromatin binding innate immune response positive regulation of transcription, DNA-templated negative regulation of ubiquitin-protein transferase activity defense response to virus protein autoubiquitination protein K48-linked ubiquitination DNA repair-dependent chromatin remodeling positive regulation of protein localization to nucleus positive regulation of protein localization to early endosome positive regulation of receptor catabolic process
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline