    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • XPO6 Antibodies
          Bethyl Laboratories
          XPO6/Exportin 6 Polyclonal Antibody
          View all (6) XPO6 antibodies
          Tech Support
          Cite XPO6/Exportin 6 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          XPO6/Exportin 6 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          XPO6/Exportin 6 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          XPO6/Exportin 6 Antibody (A301-205A-T) in IP
          Detection of human XPO6/Exportin 6 by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HeLa cells. Antibodies: Affinity purified rabbit anti-XPO6/Exportin 6 antibody (Product # A301-205A) used for WB at 1 µg/mL used for IP at 3 µg/mg lysate. XPO6/Exportin 6 was also immunoprecipitated by rabbit anti-XPO6/Exportin 6 antibody BL4796, which recognizes an upstr... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          XPO6/Exportin 6 Antibody in Immunoprecipitation (IP)
          XPO6/Exportin 6 Polyclonal Antibody
          Product Details
          A301-205A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-5 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1075 and 1125 of Human exportin 6.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Mouse and X. laevis
          Target Information
          The protein encoded by this gene is a member of the importin-beta family. Members of this family are regulated by the GTPase Ran to mediate transport of cargo across the nuclear envelope. This protein has been shown to mediate nuclear export of profilin-actin complexes. A pseudogene of this gene is located on the long arm of chromosome 14. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
          Bioinformatics
          Protein Aliases: Exp6; Exportin-6; Ran-binding protein 20; unnamed protein product
          View more View less
          Gene Aliases: EXP6; KIAA0370; RANBP20; XPO6
          View more View less
          UniProt ID: (Human) Q96QU8
          View more View less
          Entrez Gene ID: (Human) 23214
          View more View less
          Function(s)
          nuclear export signal receptor activity protein binding small GTPase binding
          Process(es)
          protein export from nucleus intracellular protein transport protein transport
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline