    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • EEF2K Antibodies
          Bethyl Laboratories
          eEF2 Kinase Polyclonal Antibody
          View all (25) EEF2K antibodies
          Tech Support
          Cite eEF2 Kinase Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          eEF2 Kinase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          eEF2 Kinase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          eEF2 Kinase Antibody (A301-686A-T) in IHC (P)
          Detection of human eEF2 Kinase by immunohistochemistry. Sample: FFPE section of human ovarian carcinoma. Antibody: Affinity purified rabbit anti-eEF2 Kinase (Product # A301-686A) used at a dilution of 1:1,000 (1 µg/mL). Detection: DAB. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          eEF2 Kinase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          eEF2 Kinase Antibody in Western Blot (WB)
          eEF2 Kinase Polyclonal Antibody
          Product Details
          A301-686A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunohistochemistry (IHC)
          1:500-1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500-1:1,000
          -
          Immunoprecipitation (IP)
          2-5 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1 and 50 of human calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
          Based on 100% sequence identity, this antibody is predicted to react with Rat
          Target Information
          EEF2K is a highly conserved Serine/Threoning kinase in the calmodulin signaling pathway whose activity is increased in cancers including breast cancer. EEF2K (eukaryotic elongation factor 2 kinase) is a threonine kinase that regulates protein synthesis by controlling the rate of peptide chain elongation. Up activation, by a variety of upstream kinases including AMPK or TRPM7, EEF2K phosphorylates the elongation factor EEF2 at a single site. Then, EEF2K renders EEF2 unable to bind ribosomes and inactive - in turn, the rate of protein synthesis is reduced.
          Bioinformatics
          Protein Aliases: alternative protein EEF2K; Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase; calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase; calmodulin-dependent protein kinase III; EC 2.7.11.20; eEF-2 kinase; EF2K; elongation factor-2 kinase; euk; eukaroytic elongation factor 2 kinase; Eukaryotic elongation factor 2 kinase; kinase eEF2K
          View more View less
          Gene Aliases: CaMKIII; eEF-2K; EEF2K; HSU93850
          View more View less
          UniProt ID: (Human) O00418
          View more View less
          Entrez Gene ID: (Human) 29904
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity elongation factor-2 kinase activity calcium ion binding calmodulin binding ATP binding translation factor activity, RNA binding kinase activity transferase activity
          Process(es)
          response to ischemia translational elongation myosin II filament disassembly cellular response to insulin stimulus negative regulation of apoptotic process positive regulation of endocytosis protein autophosphorylation positive regulation of synapse assembly positive regulation of dendritic spine morphogenesis cellular response to calcium ion cellular response to cAMP cellular response to anoxia regulation of translation at postsynapse cellular response to brain-derived neurotrophic factor stimulus response to prolactin
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline