    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CEP135 Antibodies
          Bethyl Laboratories
          CEP135 Polyclonal Antibody
          View all (8) CEP135 antibodies
          Tech Support
          Cite CEP135 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CEP135 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          CEP135 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CEP135 Antibody (A302-250A-T) in IP
          Detection of human CEP135 by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HeLa cells. Antibodies: Affinity purified rabbit anti-CEP135 antibody (Product # A302-250A) used for IP at 10 µg/mg lysate. CEP135 was also immunoprecipitated by rabbit anti-CEP135 antibody A302-251A, which recognizes a downstream epitope. For blotting immunoprecipitated CEP135, an antibody (BL6907) to an upstream epitope was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CEP135 Antibody in Immunoprecipitation (IP)
          CEP135 Polyclonal Antibody
          Product Details
          A302-250A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          5-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 900 and 950 of human centrosomal protein 135kDa.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          LATS1 is a putative serine/threonine kinase that localizes to the mitotic apparatus and complexes with cell cycle controller CDC2 kinase in early mitosis. The protein is phosphorylated in a cell-cycle dependent manner, with late prophase phosphorylation remaining through metaphase. The N-terminal region of the protein binds CDC2 to form a complex showing reduced H1 histone kinase activity, indicating a role as a negative regulator of CDC2/cyclin A. In addition, the C-terminal kinase domain binds to its own N-terminal region, suggesting potential negative regulation through interference with complex formation via intramolecular binding. Biochemical and genetic data suggest a role as a tumor suppressor. This is supported by studies in knockout mice showing development of soft-tissue sarcomas, ovarian stromal cell tumors and a high sensitivity to carcinogenic treatments.
          Bioinformatics
          Protein Aliases: centrosomal protein 135kDa; Centrosomal protein 4; Centrosomal protein of 135 kDa; centrosome protein cep135; Cep135
          View more View less
          Gene Aliases: CEP135; CEP4; KIAA0635; MCPH8
          View more View less
          UniProt ID: (Human) Q66GS9
          View more View less
          Entrez Gene ID: (Human) 9662
          View more View less
          Function(s)
          protein binding microtubule binding tubulin binding identical protein binding protein homodimerization activity
          Process(es)
          centriole replication centriole-centriole cohesion positive regulation of nonmotile primary cilium assembly positive regulation of establishment of protein localization
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline