    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cyclin Y Antibodies
          Bethyl Laboratories
          Cyclin Y Polyclonal Antibody
          View all (15) Cyclin Y antibodies
          Tech Support
          Cite Cyclin Y Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Cyclin Y Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Cyclin Y Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Cyclin Y Antibody (A302-376A-T) in WB
          Detection of human Cyclin Y by western blot. Samples: Whole cell lysate (50 µg) from HeLa (H), HEK293T (T), mouse NIH 3T3 (3), and MCF-7 (M) cells. Antibodies: Affinity purified rabbit anti-Cyclin Y antibody (Product # A302-376A) used for WB at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cyclin Y Antibody in Western Blot (WB)
          Cyclin Y Polyclonal Antibody
          Product Details
          A302-376A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,500
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 291 and 341 of human cyclin Y.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
          Bioinformatics
          Protein Aliases: CBCP1; Cyc-Y; Cyclin box protein 1; Cyclin fold protein 1; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; unnamed protein product
          View more View less
          Gene Aliases: C10orf9; CBCP1; CCNX; CCNY; CFP1
          View more View less
          UniProt ID: (Human) Q8ND76
          View more View less
          Entrez Gene ID: (Human) 219771
          View more View less
          Function(s)
          protein binding cyclin-dependent protein serine/threonine kinase regulator activity protein kinase binding
          Process(es)
          G2/M transition of mitotic cell cycle spermatogenesis positive regulation of autophagy Wnt signaling pathway positive regulation of cyclin-dependent protein serine/threonine kinase activity cell division regulation of canonical Wnt signaling pathway
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline