    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BEGAIN Antibodies
          Bethyl Laboratories
          BEGAIN Polyclonal Antibody
          View all (25) BEGAIN antibodies
          Tech Support
          Cite BEGAIN Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          BEGAIN Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          BEGAIN Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          BEGAIN Antibody (A302-792A-T) in IP
          Detection of human BEGAIN by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP, 20% of IP loaded) from HeLa cells. Antibodies: Affinity purified rabbit anti-BEGAIN antibody (Product # A302-792A) used for IP at 10 µg/mg lysate. BEGAIN was also immunoprecipitated by rabbit anti-BEGAIN antibody A302-793A, which recognizes a downstream epitope. For blotting immunoprecipitated BEGAIN, A302-793A was used at 1 µg/mL,. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BEGAIN Antibody in Immunoprecipitation (IP)
          BEGAIN Polyclonal Antibody
          Product Details
          A302-792A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          5-15 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 493 and 543 of human Brain-Enriched Guanylate Kinase-Associated Protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          May sustain the structure of the postsynaptic density (PSD). Target protein binds the guanylate kinase domain of PSD-95/SAP90, may play a role in the post-synaptic membrane.
          Bioinformatics
          Protein Aliases: brain-enriched guanylate kinase-associated homolog; Brain-enriched guanylate kinase-associated protein
          View more View less
          Gene Aliases: BEGAIN; KIAA1446
          View more View less
          UniProt ID: (Human) Q9BUH8
          View more View less
          Entrez Gene ID: (Human) 57596
          View more View less
          Function(s)
          protein binding
          Process(es)
          evoked excitatory postsynaptic potential
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline