    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ARHGAP11A Antibodies
          Bethyl Laboratories
          ARHGAP11A Polyclonal Antibody
          View all (6) ARHGAP11A antibodies
          Tech Support
          Cite ARHGAP11A Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          ARHGAP11A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ARHGAP11A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          ARHGAP11A Antibody (A303-097A-T) in WB
          Detection of human and mouse ARHGAP11A by western blot (h & m) and immunoprecipitation (h). Samples: Whole cell lysate from HeLa (5, 15 and 50 µg for WB; 1 mg for IP, 20% of IP loaded), HEK293T (T; 50 µg) and mouse NIH 3T3 (M; 50 µg) cells. Antibodies: Affinity purified rabbit anti-ARHGAP11A antibody (Product # A303-097A) used for WB at 0.1 µg/mL (A) and 1 µg/mL (B) and used for IP at 6 µg/mg lysate. ARHGAP11A was also immunoprecipitated by rabbit anti-ARHGAP11A antibody BL7701, which recognizes an upstream epitope. Detection: Chemilumines... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ARHGAP11A Antibody in Western Blot (WB)
          ARHGAP11A Polyclonal Antibody
          Product Details
          A303-097A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 973 and 1023 of human Rho GTPase Activating Protein 11A.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          ARHGAP11A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include signal transduction.
          Bioinformatics
          Protein Aliases: GAP (1-12); KIAA0013; Rho GTPase-activating protein 11A; Rho-type GTPase-activating protein 11A; unnamed protein product
          View more View less
          Gene Aliases: 6530401L14Rik; ARHGAP11A; GAP (1-12); KIAA0013; mKIAA0013
          View more View less
          UniProt ID: (Human) Q6P4F7, (Mouse) Q80Y19
          View more View less
          Entrez Gene ID: (Human) 9824, (Mouse) 228482
          View more View less
          Function(s)
          GTPase activator activity molecular_function G-protein modulator protein-binding activity modulator GTPase-activating protein
          Process(es)
          signal transduction positive regulation of GTPase activity regulation of small GTPase mediated signal transduction
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline