  • Applications
    • Bioprocesamiento
    • Cultivo celular y transfección
    • Terapia celular y génica
    • Cromatografía
    • Pruebas moleculares
    • Soluciones digitales
    • Extracción y análisis de ADN y ARN
    • Espectroscopía, análisis elemental y de isótopos
    • Ver todas las aplicaciones y técnicas
  • Servicios
    • Servicios 360° de CDMO y CRO
    • Servicios de CDMO
    • Servicios de CRO
    • Servicios personalizados
    • Servicios de leasing y financiación
    • Servicios de instrumentos
    • Informática de laboratorio
    • OEM y oferta comercial
    • Servicios de formación
    • Unity Lab Services
    • Ver todos los servicios
  • Ayuda y soporte técnico
    • Regístrese para obtener una cuenta
    • Cómo hacer un pedido
    • Asistencia para el instrumental
    • Centros de soporte técnico
    • Centros de formación
    • Vea todos los temas de ayuda y soporte técnico
      Cell Culture Plastics
  • A quiénes brindamos nuestros servicios
    • Industria biotecnológica
    • Sector biofarmacéutico
    • CDMO
    • Diagnósticos de laboratorio
    • Ciencias aplicadas e industriales
  • Promociones
  • Contacto
  • Pedido rápido
  • Estado del pedido y seguimiento
  • Documentos y certificados
Thermo Fisher Scientific Logo
Search
Buscar
Search button
          • Estado del pedido
          • Pedido rápido
            • Cuenta
            • Pedidos
            • Connect: laboratorio, datos, aplicaciones
            • Productos y proyectos personalizados
            • Services Central
          • Primary Antibodies ›
          • TSC22D1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Bethyl Laboratories
          TSC22D1 Polyclonal Antibody
          View all (38) TSC22D1 antibodies
          Tech Support
          Cite TSC22D1 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          TSC22D1 Polyclonal Antibody
          Product Details
          A303-582A-M
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunohistochemistry (IHC)
          1:500-1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500-1:2,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1023 and 1073 of Human TSC22 (TGFB-stimulated clone 22 ) domain family, member 1.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
          Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
          Target Information
          TSC22 domain family protein, or TSC22, is a transcription factor that belongs to the large family of early response genes. This transcriptional repressor is known to act on the C-type natriuretic peptide (CNP) promoter. TSC22 belongs to the TSC-22/Dip/Bun family and is an intracellular protein that may be found in the cytoplasm or nucleus. This transcription factor is known to regulate cell growth, differentiation and cell death and is involved in modulating the transcriptional activity of Smad3 and Smad4. This protein is ubiquitously expressed in most tissues and widely in both fetal and adult tissues. It is generally expressed in aortic endothelial cells, and induced by cytokines, including TGFB. These proteins may be possible therapeutic targets of leukemia and prostate cancer.
          Pedidos Plus Icon Minus Icon
          • Estado del pedido
          • Ayuda con el pedido
          • Pedido rápido
          • Centro de suministros
          • eProcurement (B2B)
          Asistencia Plus Icon Minus Icon
          • Ayuda y soporte técnico
          • Contacto
          • Centros de soporte técnico
          • Documentos y certificados
          • Informar de un problema del sitio web
          Recursos Plus Icon Minus Icon
          • Centros de formación
          • Promociones
          • Eventos y seminarios web
          • Redes sociales
          Acerca de Thermo Fisher Plus Icon Minus Icon
          • Acerca de nosotros Acerca de nosotros
          • Carreras profesionales Carreras profesionales
          • Inversores Inversores
          • Noticias Noticias
          • Responsabilidad social Responsabilidad social
          • Marcas registradas
          • Políticas y avisos
          Nuestra cartera Plus Icon Minus Icon
          Spain flag icon
          Spain
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline