FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Required for 40S ribosome biogenesis. Involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
Protein Aliases: HIV-1 rev binding protein 2; HIV-1 Rev-binding protein 2; KRR-R motif-containing protein 1; KRR1 small subunit processome component homolog; KRR1, small subunit (SSU) processome component, homolog; Rev interacting protein; Rev-interacting protein 1; Rip-1; unnamed protein product
Gene Aliases: HRB2; KRR1; RIP-1
UniProt ID: (Human) Q13601
Entrez Gene ID: (Human) 11103