    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TXLNG Antibodies
          Bethyl Laboratories
          Gamma-Taxilin Polyclonal Antibody
          View all (8) TXLNG antibodies
          Tech Support
          Cite Gamma-Taxilin Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Gamma-Taxilin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Gamma-Taxilin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Gamma-Taxilin Antibody (A304-041A-T) in WB
          Detection of human Gamma-Taxilin by western blot. Samples: Whole cell lysate (50 µg) from HEK293T, HeLa, and Jurkat cells. Antibodies: Affinity purified rabbit anti-Gamma-Taxilin antibody (Product # A304-041A) (lot A304-041A-1) used for WB at 0.04 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Gamma-Taxilin Antibody in Western Blot (WB)
          Gamma-Taxilin Antibody in Immunoprecipitation (IP)
          Gamma-Taxilin Polyclonal Antibody
          Product Details
          A304-041A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:25,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 25 to 75 of human Taxilin Gamma.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          TXLNG gene ontology annotations related to this gene include regulation of bone mineralization; regulation of cell cycle process.
          Bioinformatics
          Protein Aliases: Environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; Factor inhibiting ATF4-mediated transcription; FIAT; Gamma-taxilin; lipopolysaccharide specific response-5 protein; Lipopolysaccharide-specific response protein 5; LSR-5; unnamed protein product
          View more View less
          Gene Aliases: CXorf15; ELRG; FIAT; LSR5; TXLNG; TXLNGX
          View more View less
          UniProt ID: (Human) Q9NUQ3
          View more View less
          Entrez Gene ID: (Human) 55787
          View more View less
          Function(s)
          syntaxin binding DNA-binding transcription factor binding
          Process(es)
          regulation of cell cycle process regulation of bone mineralization regulation of cell cycle
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline