    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DNAJC1 Antibodies
          Bethyl Laboratories
          MTJ1 Polyclonal Antibody
          View all (6) DNAJC1 antibodies
          Tech Support
          Cite MTJ1 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MTJ1 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          MTJ1 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MTJ1 Antibody (A304-056A-T) in IP
          Detection of human MTJ1 by western blot of immunoprecipitates. Samples: Whole cell lysate (1 mg for IP; 20% of IP loaded) from Jurkat cells. Antibodies: Affinity purified rabbit anti-MTJ1 antibody (Product # A304-056A) (lot 1) used for IP at 6 µg/mg lysate. MTJ1 was also immunoprecipitated by rabbit anti-MTJ1 antibody A304-055A. For blotting immunoprecipitated MTJ1, A304-055A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 10 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MTJ1 Antibody in Immunoprecipitation (IP)
          MTJ1 Polyclonal Antibody
          Product Details
          A304-056A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 504 to 554 of human DnaJ (Hsp40) Homolog, Subfamily C, Member 1.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Mouse
          Target Information
          The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
          Bioinformatics
          Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 1; DnaJ homolog subfamily C member 1; DnaJ protein homolog MTJ1; DnaJ-like protein; unnamed protein product
          View more View less
          Gene Aliases: DNAJC1; DNAJL1; ERdj1; HTJ1; MTJ1
          View more View less
          UniProt ID: (Human) Q96KC8
          View more View less
          Entrez Gene ID: (Human) 64215
          View more View less
          Function(s)
          ATPase activator activity DNA binding protein binding chaperone binding
          Process(es)
          regulation of translation protein folding negative regulation of proteolysis regulation of protein secretion regulation of protein metabolic process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline