    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MYOF Antibodies
          Bethyl Laboratories
          Myoferlin Polyclonal Antibody
          View all (11) MYOF antibodies
          Tech Support
          Cite Myoferlin Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Myoferlin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Myoferlin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Myoferlin Antibody (A304-387A-T) in WB
          Detection of human Myoferlin by western blot. Samples: Whole cell lysate (50 µg) prepared using NETN buffer from HeLa cells. Antibodies: Affinity purified rabbit anti-Myoferlin antibody (Product # A304-387A) (lot A304-387A-1) used for WB at 0.4 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Myoferlin Antibody in Western Blot (WB)
          Myoferlin Antibody in Immunoprecipitation (IP)
          Myoferlin Polyclonal Antibody
          Product Details
          A304-387A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:5,000
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 700 to 750 of human Myoferlin.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined.
          Bioinformatics
          Protein Aliases: fer-1-like 3, myoferlin; fer-1-like family member 3; Fer-1-like protein 3; ferlin; ferlin family member; Myoferlin; unnamed protein product
          View more View less
          Gene Aliases: FER1L3; HAE7; KIAA1207; MYOF
          View more View less
          UniProt ID: (Human) Q9NZM1
          View more View less
          Entrez Gene ID: (Human) 26509
          View more View less
          Function(s)
          calcium ion binding protein binding phospholipid binding metal ion binding
          Process(es)
          plasma membrane repair muscle contraction blood circulation regulation of neurotransmitter secretion
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline