    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PKP4 Antibodies
          Bethyl Laboratories
          p0071 Polyclonal Antibody
          View all (15) PKP4 antibodies
          Tech Support
          Cite p0071 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          p0071 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          p0071 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          p0071 Antibody (A304-649A-T) in WB
          Detection of human p0071 by western blot. Samples: Whole cell lysate (50 µg) from HeLa, HEK293T, Jurkat and mouse TCMK-1 cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-p0071 antibody (Product # A304-649A) (lot A304-649A-1) used for WB at 0.1 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          p0071 Antibody in Western Blot (WB)
          p0071 Polyclonal Antibody
          Product Details
          A304-649A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1 to 50 of human Plakophilin 4.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          Plays a role as a regulator of Rho activity during cytokinesis. May play a role in junctional plaques.
          Bioinformatics
          Protein Aliases: catenin 4; p0071; Plakophilin-4; similar to p0071 protein encoded by GenBank Accession Number X81889; armadillo repeat-containing protein similar to GT24 p120(cas), to beta catenin and to armadillo
          View more View less
          Gene Aliases: p0071; PKP4
          View more View less
          UniProt ID: (Human) Q99569
          View more View less
          Entrez Gene ID: (Human) 8502
          View more View less
          Function(s)
          protein binding cadherin binding
          Process(es)
          cell-cell junction assembly cell adhesion cell-cell signaling regulation of cell adhesion positive regulation of cytokinesis regulation of Rho protein signal transduction cell-cell adhesion
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline