    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • TEX2 Antibodies
          Bethyl Laboratories
          TEX2 Polyclonal Antibody
          View all (5) TEX2 antibodies
          Tech Support
          Cite TEX2 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TEX2 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          TEX2 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TEX2 Antibody (A304-705A-T) in IP
          Detection of human TEX2 by western blot of immunoprecipitates. Samples: Whole cell lysate (0.5 or 1.0 mg per IP reaction; 20% of IP loaded) from Jurkat cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-TEX2 antibody (Product # A304-705A) (lot 1) used for IP at 6 µg per reaction. TEX2 was also immunoprecipitated by rabbit anti-TEX2 antibody BL18225. For blotting immunoprecipitated TEX2, A304-705A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TEX2 Antibody in Immunoprecipitation (IP)
          TEX2 Polyclonal Antibody
          Product Details
          A304-705A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 350 to 400 of Human Testis Expressed 2.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          TEX2 is a multi-pass membrane protein. The exact function of TEX2 remains unknown.
          Bioinformatics
          Protein Aliases: testis expressed sequence 2; Testis-expressed protein 2; testis-expressed sequence 2 protein; Transmembrane protein 96; unnamed protein product
          View more View less
          Gene Aliases: HT008; KIAA1738; TEX2; TMEM96
          View more View less
          UniProt ID: (Human) Q8IWB9
          View more View less
          Entrez Gene ID: (Human) 55852
          View more View less
          Function(s)
          lipid binding
          Process(es)
          sphingolipid metabolic process lipid transport signal transduction
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline