    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RPL21 Antibodies
          Bethyl Laboratories
          RPL21/Ribosomal Protein L21 Polyclonal Antibody
          View all (7) RPL21 antibodies
          Tech Support
          Cite RPL21/Ribosomal Protein L21 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          RPL21/Ribosomal Protein L21 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RPL21/Ribosomal Protein L21 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          RPL21/Ribosomal Protein L21 Antibody (A305-031A-T) in WB
          Detection of human and mouse RPL21 by western blot. Samples: Whole cell lysate (50 µg) from HeLa, HEK293T, Jurkat, mouse TCMK-1, and mouse NIH 3T3 cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-RPL21 antibody (Product # A305-031A) (lot A305-031A-1) used for WB at 0.1 µg/mL. Detection: Chemiluminescence with an exposure time of 10 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RPL21/Ribosomal Protein L21 Antibody in Western Blot (WB)
          RPL21/Ribosomal Protein L21 Polyclonal Antibody
          Product Details
          A305-031A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1 to 50 of Human 60S ribosomal protein L21.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Rat, Goat and Pig
          Target Information
          This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: 60S ribosomal protein L21; Large ribosomal subunit protein eL21; RPL21; unnamed protein product
          View more View less
          Gene Aliases: 8430440E03Rik; eL21; HYPT12; L21; RPL21
          View more View less
          UniProt ID: (Human) P46778, (Mouse) O09167
          View more View less
          Entrez Gene ID: (Human) 6144, (Mouse) 19933
          View more View less
          Function(s)
          RNA binding structural constituent of ribosome protein binding poly(A) RNA binding ribosomal protein
          Process(es)
          cytoplasmic translation translation
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline