    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ETEA Antibodies
          Bethyl Laboratories
          FAF2/ETEA Polyclonal Antibody
          View all (16) ETEA antibodies
          Tech Support
          Cite FAF2/ETEA Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          FAF2/ETEA Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FAF2/ETEA Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          FAF2/ETEA Antibody (A305-378A-T) in IP
          Detection of human FAF2 by western blot of immunoprecipitates. Samples: Whole cell lysate (0.5 or 1.0 mg per IP reaction; 20% of IP loaded) from 293T cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-FAF2 antibody (Product # A305-378A) (lot 1) used for IP at 6 µg per reaction. FAF2 was also immunoprecipitated by rabbit anti-FAF2 antibody A305-379A. For blotting immunoprecipitated FAF2, A305-378A was used at 0.4 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FAF2/ETEA Antibody in Immunoprecipitation (IP)
          FAF2/ETEA Polyclonal Antibody
          Product Details
          A305-378A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 350 to 400 of human FAS-associated factor 2.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          phosphate/tris citrate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
          Target Information
          The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
          Bioinformatics
          Protein Aliases: expressed in T-cells and eosinophils in atopic dermatitis; FAS-associated factor 2; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8; unnamed protein product
          View more View less
          Gene Aliases: ETEA; FAF2; KIAA0887; UBXD8; UBXN3B
          View more View less
          UniProt ID: (Human) Q96CS3
          View more View less
          Entrez Gene ID: (Human) 23197
          View more View less
          Function(s)
          protein binding protein binding, bridging ubiquitin protein ligase binding lipase binding ubiquitin binding lipase inhibitor activity
          Process(es)
          response to unfolded protein proteasomal protein catabolic process retrograde protein transport, ER to cytosol lipid particle organization stress granule disassembly ERAD pathway proteasome-mediated ubiquitin-dependent protein catabolic process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline