    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • LAG-3 (CD223) Antibodies
          Bethyl Laboratories
          LAG3 Recombinant Rabbit Monoclonal Antibody (BLR028F)
          View all (126) LAG-3 (CD223) antibodies
          Tech Support
          Cite LAG3 Recombinant Rabbit Monoclonal Antibody (BLR028F)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          LAG3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LAG3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          LAG3 Antibody (A700-028-T) in ICC/IF
          Detection of human LAG3 by immunocytochemistry. Sample: FFPE section of HDLM-2 cells. Antibody: Rabbit anti-LAG3 recombinant monoclonal antibody [BLR028F] (Product # A700-028) (lot 2). Secondary: HRP-conjugated goat anti-rabbit IgG (A120-501P). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          LAG3 Antibody in Immunocytochemistry (ICC/IF)
          LAG3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LAG3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LAG3 Antibody in Western Blot (WB)
          LAG3 Antibody in Flow Cytometry (Flow)
          LAG3 Antibody in Immunoprecipitation (IP)
          LAG3 Recombinant Rabbit Monoclonal Antibody (BLR028F)
          Product Details
          A700-028-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:100-1:500
          -
          Flow Cytometry (Flow)
          1 µL per 1 x 10^6 cells
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          BLR028F
          Immunogen
          Recombinant protein representing the extracellular domain of human Lymphocyte activation gene 3 (LAG3, residues 23-450).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          BBS, pH 8.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
          Target Information
          LAG-3 is a 70-kDa surface glycoprotein in the immunoglobulin superfamily, with structural homology to CD4. It binds to MHC class II molecules with higher affinity than CD4 and is involved in the negative regulation of T cell activation and homeostatic proliferation. LAG-3 is expressed on the surface of activated T cells, including regulatory T cells, and NK cells. Notably, CD8+ T cells express LAG-3 at significantly higher levels than CD4+ T cells. Additionally, the coexpression of LAG-3 and CD49b has been proposed as a marker for identifying Type 1 regulatory T cells (Tr1 cells) in both humans and mice. This highlights LAG-3s role in immune regulation and its potential as a target for therapeutic interventions in immune-related conditions.
          Bioinformatics
          Protein Aliases: CD223; LAG-3; Lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; sLAG 3; sLAG3; soluble LAG 3lymphocyte activating 3; soluble LAG3; unnamed protein product
          View more View less
          Gene Aliases: CD223; FDC; LAG3
          View more View less
          UniProt ID: (Human) P18627
          View more View less
          Entrez Gene ID: (Human) 3902
          View more View less
          Function(s)
          antigen binding transmembrane signaling receptor activity protein binding MHC class II protein binding
          Process(es)
          adaptive immune response plasmacytoid dendritic cell activation immune system process cell surface receptor signaling pathway negative regulation of interleukin-2 production T cell activation natural killer cell mediated cytotoxicity innate immune response negative regulation of regulatory T cell differentiation positive regulation of natural killer cell mediated cytotoxicity regulation of immune response negative regulation of T cell activation
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline