FIGURE: 1 / 4
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Essential component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8A constitutes a pore-forming subunit or whether it is closely associated with the pore and mediates channel properties such as ion selectivity. Involved in B-cell development: required for the pro-B cell to pre-B cell transition. Also required for T-cell development.
Protein Aliases: ebo; HsLRRC8A; leucine rich repeat containing 8 family member A; leucine rich repeat containing 8A; leucine-rich repeat-containing 8; Leucine-rich repeat-containing protein 8A; Protein ebouriffe; Swelling protein 1; Swelling protein 1 SWELL1; unnamed protein product; Volume-regulated anion channel subunit LRRC8A
Gene Aliases: AGM5; HsLRRC8A; KIAA1437; LRRC8; LRRC8A; mKIAA1437; SWELL1; UNQ221/PRO247
UniProt ID: (Human) Q8IWT6, (Mouse) Q80WG5, (Rat) Q4V8I7
Entrez Gene ID: (Human) 56262, (Mouse) 241296, (Rat) 311846