Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • alanyl-tRNA Synthetase Antibodies

          Abnova

          AARS Polyclonal Antibody, MaxPab™

          View all (11) alanyl-tRNA Synthetase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AARS Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          AARS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          AARS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AARS Antibody (H00000016-D01P) in WB

          AARS MaxPab rabbit polyclonal antibody. Western Blot analysis of AARS expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AARS Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          AARS Polyclonal Antibody, MaxPab™

          Product Details

          H00000016-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AARS (NP_001596.2, 1 a.a. approximately 968 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDSTLTASEI RQRFIDFFKR NEHTYVHSSA TIPLDDPTLL FANAGMNQFK PIFLNTIDPS HPMAKLSRAA NTQKCIRAGG KHNDLDDVGK DVYHHTFFEM LGSWSFGDYF KELACKMALE LLTQEFGIPI ERLYVTYFGG DEAAGLEADL ECKQIWQNLG LDDTKILPGN MKDNFWEMGD TGPCGPCSEI HYDRIGGRDA AHLVNQDDPN VLEIWNLVFI QYNREADGIL KPLPKKSIDT GMGLERLVSV LQNKMSNYDT DLFVPYFEAI QKGTGARPYT GKVGAEDADG IDMAYRVLAD HARTITVALA DGGRPDNTGR GYVLRRILRR AVRYAHEKLN ASRGFFATLV DVVVQSLGDA FPELKKDPDM VKDIINEEEV QFLKTLSRGR RILDRKIQSL GDSKTIPGDT AWLLYDTYGF PVDLTGLIAE EKGLVVDMDG FEEERKLAQL KSQGKGAGGE DLIMLDIYAI EELRARGLEV TDDSPKYNYH LDSSGSYVFE NTVATVMALR REKMFVEEVS TGQECGVVLD KTCFYAEQGG QIYDEGYLVK VDDSSEDKTE FTVKNAQVRG GYVLHIGTIY GDLKVGDQVW LFIDEPRRRP IMSNHTATHI LNFALRSVLG EADQKGSLVA PDRLRFDFTA KGAMSTQQIK KAEEIANEMI EAAKAVYTQD CPLAAAKAIQ GLRAVFDETY PDPVRVVSIG VPVSELLDDP SGPAGSLTSV EFCGGTHLRN SSHAGAFVIV TEEAIAKGIR RIVAVTGAEA QKALRKAESL KKCLSVMEAK VKAQTAPNKD VQREIADLGE ALATAVIPQW QKDELRETLK SLKKVMDDLD RASKADVQKR VLEKTKQFID SNPNQPLVIL EMESGASAKA LNEALKLFKM HSPQTSAMLF TVDNEAGKIT CLCQVPQNAA NRGLKASEWV QQVSGLMDGK GGGKDVSAQA TGKNVGCLQE ALQLATSFAQ LRLGDVKN

          Target Information

          The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E. coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alanine tRNA ligase 1, cytoplasmic; alanyl-tRNA synthetase, cytoplasmic; alaRS; protein lactyltransferase AARS1; renal carcinoma antigen NY-REN-42; unnamed protein product

          View more View less

          Gene Aliases: AARS; CMT2N; DEE29; EIEE29; HDLS2; TTD8

          View more View less

          UniProt ID: (Human) P49588

          View more View less

          Entrez Gene ID: (Human) 16

          View more View less

          Function(s)
          tRNA binding nucleotide binding aminoacyl-tRNA editing activity Ser-tRNA(Ala) hydrolase activity nucleic acid binding aminoacyl-tRNA ligase activity alanine-tRNA ligase activity ATP binding zinc ion binding amino acid binding peptide lactyltransferase (ATP-dependent) activity
          Process(es)
          tRNA modification tRNA aminoacylation for protein translation alanyl-tRNA aminoacylation tRNA processing positive regulation of hippo signaling tRNA aminoacylation aminoacyl-tRNA metabolism involved in translational fidelity regulation of cytoplasmic translational fidelity negative regulation of signal transduction by p53 class mediator
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch