Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: DITMVICPG IANHSEKQYI RTFVDYAQKN GYRCAVLNHL GALPNIELTS PRMFTYGCTW EFGAMVNYIK KTYPLTQLVV VGFSLGGNIV CKYLGETQAN QEKVLCCVSV CQGYSALRAQ ETFMQWDQCQ RFYNFLMADN MKKIILSHRQ ALFGDHVKKP QSLEDTDLSR LYTATSLMQI DDNVMRKFHG YNSLKEYYEE ESCMRYLHRI YVPLMLVNAA DDPLVHESLL TIPKSLSEKR ENVMFVLPLH GGHLGFFEGS VLFPEPLTWM DKLVVEYANA ICQWERNKLQ CSDTEQVEAD LE (125-425 aa encoded by BC019248)
This gene encodes a protein containing an alpha/beta hydrolase fold, which is a catalytic domain found in a very wide range of enzymes. The function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-arachidonoylglycerol hydrolase; abhydrolase domain-containing protein 2; acetylesterase; alpha/beta hydrolase domain containing protein 2; alpha/beta hydrolase fold protein; lung alpha/beta hydrolase 2; mmLABH2; progesterone-sensitive lipase; protein PHPS1-2; testicular tissue protein Li 6; triacylglycerol lipase; unnamed protein product
Gene Aliases: 2210009N18Rik; HS1-2; Labh-2; LABH2; PHPS1-2
UniProt ID: (Human) P08910, (Mouse) Q9QXM0
Entrez Gene ID: (Human) 11057, (Rat) 293050, (Mouse) 54608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support