Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • Abl2 Antibodies

          Abnova

          ABL2 Polyclonal Antibody, MaxPab™

          View all (21) Abl2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ABL2 Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          ABL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ABL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ABL2 Antibody (H00000027-B01P) in WB

          Western Blot analysis of ABL2 expression in transfected 293T cell line by ABL2 MaxPab polyclonal antibody. Lane 1: ABL2 transfected lysate(126.70 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ABL2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          ABL2 Polyclonal Antibody, MaxPab™

          Product Details

          H00000027-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ABL2 (AAH65912.1, 1 a.a. approximately 1167 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MVLGTVLLPP NSYGRDQDTS LCCLCTEASE SALPDLTDHF ASCVEDGFEG DKTGGSSPEA LHRPYGCDVE PQALNEAIRW SSKENLLGAT ESDPNLFVAL YDFVASGDNT LSITKGEKLR VLGYNQNGEW SEVRSKNGQG WVPSNYITPV NSLEKHSWYH GPVSRSAAEY LLSSLINGSF LVRESESSPG QLSISLRYEG RVYHYRINTT ADGKVYVTAE SRFSTLAELV HHHSTVADGL VTTLHYPAPK CNKPTVYGVS PIHDKWEMER TDITMKHKLG GGQYGEVYVG VWKKYSLTVA VKTLKEDTME VEEFLKEAAV MKEIKHPNLV QLLGVCTLEP PFYIVTEYMP YGNLLDYLRE CNREEVTAVV LLYMATQISS AMEYLEKKNF IHRDLAARNC LVGENHVVKV ADFGLSRLMT GDTYTAHAGA KFPIKWTAPE SLAYNTFSIK SDVWAFGVLL WEIATYGMSP YPGIDLSQVY DLLEKGYRME QPEGCPPKVY ELMRACWKWS PADRPSFAET HQAFETMFHD SSISEEVAEE LGRAASSSSV VPYLPRLPIL PSKTRTLKKQ VENKENIEGA QDATENSASS LAPGFIRGAQ ASSGSPALPR KQRDKSPSSL LEDAKETCFT RDRKGGFFSS FMKKRNAPTP PKRSSSFREM ENQPHKKYEL TGNFSSVASL QHADGFSFTP AQQEANLVPP KCYGGSFAQR NLCNDDGGGG GGSGTAGGGW SGITGFFTPR LIKKTLGLRA GKPTASDDTS KPFPRSNSTS SMSSGLPEQD RMAMTLPRNC QRSKLQLERT VSTSSQPEEN VDRANDMLPK KSEESAAPSR ERPKAKLLPR GATALPLRTP SGDLAITEKD PPGVGVAGVA AAPKGKEKNG GARLGMAGVP EDGEQPGWPS PAKAAPVLPT THNHKVPVLI SPTLKHTPAD VQLIGTDSQG NKFKLLSEHQ VTSSGDKDRP RRVKPKCAPP PPPVMRLLQH PSICSDPTEE PTALTAGQST SETQEGGKKA ALGAVPISGK AGRPVMPPPQ VPLPTSSISP AKMANGTAGT KVALRKTKQA AEKISADKIS KEALLECADL LSSALTEPVP NSQLVDTGHQ LLDYCSGYVD CIPQTRNKFA FREAVSKLEL SLQELQVSSA AAGVPGTNPV LNNLLSCVQE ISDVVQR

          Target Information

          ABL2 (Arg) is an Abl-related non-receptor protein tyrosine kinase that links cell surface receptors to cell proliferation and/or cytoskeletal reorganization. Abl2 encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. Abl2 is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Abelson tyrosine-protein kinase 2; abelson-related gene protein; arg tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; FLJ22224; FLJ31718; FLJ41441; tyrosine-protein kinase ARG; v-abl Abelson murine leukemia viral oncogene homolog 2

          View more View less

          Gene Aliases: ABLL; ARG

          View more View less

          UniProt ID: (Human) P42684

          View more View less

          Entrez Gene ID: (Human) 27

          View more View less

          Function(s)
          magnesium ion binding phosphotyrosine binding actin monomer binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity protein binding ATP binding enzyme activator activity enzyme binding manganese ion binding actin filament binding
          Process(es)
          signal transduction epidermal growth factor receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration regulation of autophagy positive regulation of neuron projection development peptidyl-tyrosine phosphorylation regulation of endocytosis regulation of cell adhesion regulation of actin cytoskeleton organization cellular response to oxidative stress negative regulation of Rho protein signal transduction exploration behavior protein modification process cellular response to retinoic acid positive regulation of establishment of T cell polarity regulation of cell motility positive regulation of T cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-5d76f9f486-9kkwj:80/100.66.77.245:80.
          git-commit: 08b979eaab39376f31a39358ffa32de998e3c7f7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.6.0-offline-prod