Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • ABO Antibodies

          Invitrogen

          ABO Polyclonal Antibody

          View all (9) ABO antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ABO Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          ABO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ABO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ABO Antibody (PA5-37352) in IHC (P)

          Immunohistochemistry analysis of ABO in paraffin-embedded human esophageal cancer tissue. Samples were incubated with ABO polyclonal antibody (Product # PA5-37352). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ABO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ABO Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          ABO Polyclonal Antibody

          Product Details

          PA5-37352

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant full length human ABO
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2554023

          Product Specific Information

          This antibody detects endogenous protein at a molecular weight of 41 kDa.

          Purity is >95% by SDS-PAGE.

          Target Information

          Histo-blood group ABO system transferase (NAGAT) is a member of the glycosyltransferase 6 family of proteins and the basis of the ABO blood group system. The histo-blood group ABO involves three carbohydrate antigens: A, B and H; the NAGAT protein functions as the basis of this group. A, B and AB individuals express a glycosyltransferase activity converting the H antigen to the A antigen (by addition of UDP-GalNAc) or to the B antigen (by addition of UDP-Gal), while O individuals do not express this glycosyltransferase activity. The B phenotype of NAGAT differs from the A form by a few residue substitutions, whereas the O form is a result of a single base frame-shift deletion in the N-terminal extremity of the gene. The NAGAT protein localizes to the Golgi apparatus and its conserved DXD motif functions in cofactor binding.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: a transferase; ABO blood group (alpha 1-3-N-acetylgalactosaminyltransferase, alpha 1-3-galactosyltransferase); ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase); ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); alpha 1-3-N-acetylgalactosaminyltransferase; Any polymorphism or mutation that changes the activity or specificity of the encoded enzyme may therefore alter the ABO phenotype.; B (A) alpha 1; b blood group galactosyltransferase; b transferase; B(A) alpha-1,3-galactosyltransferase; Bel; BGAT; blood group A glycosyltransferase 2; cis-AB transferase; cis-AB transferase 2; fucosylglycoprotein 3-alpha-galactosyltransferase; fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; histo-blood group A transferase; histo-blood group A transferase (UDP-GalNAc); histo-blood group A2 transferase; histo-blood group ABO system transferase; histo-blood group B transferase; NAGAT 2; putative blood group A transferase T2; red blood cell ABO blood group B antigen 648C>G results in amino acid change of transferase; transfer products of ABO group A antigen 297A>G, 467C>T, 1003G>A lead to amino acid changes; transferase A; A glycosyltransferase; transferase B; transferase B, alpha 1-3-galactosyltransferase); Unknown; unnamed protein product

          View more View less

          Gene Aliases: A3GALNT; A3GALT1; Abo2; GTA; GTB; NAGAT

          View more View less

          UniProt ID: (Human) P16442, (Mouse) P38649, (Rat) Q8CFC4

          View more View less

          Entrez Gene ID: (Human) 28, (Mouse) 80908, (Rat) 100301568

          View more View less

          Function(s)
          nucleotide binding antigen binding glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase activity fucosylgalactoside 3-alpha-galactosyltransferase activity transferase activity, transferring hexosyl groups manganese ion binding alpha-1,3-galactosyltransferase activity UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
          Process(es)
          carbohydrate metabolic process oligosaccharide biosynthetic process positive regulation of cell proliferation response to nematode response to retinoic acid negative regulation of apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-684z6:80/100.66.78.224:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0