    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Hemopexin Antibodies
          Invitrogen
          Hemopexin Monoclonal Antibody (4), Biotin
          View all (22) Hemopexin antibodies
          Tech Support
          Cite Hemopexin Monoclonal Antibody (4), Biotin
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Hemopexin Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          Hemopexin Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Hemopexin Antibody (ABS 013-04B-01) in ELISA
          Sandwich ELISA of Hemopexin using ABS 013-32 as the capture antibody and ABS 013-04B as the biotinylated detection antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Hemopexin Antibody in ELISA (ELISA)
          Hemopexin Monoclonal Antibody (4), Biotin
          Product Details
          ABS 013-04B-01
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4
          Immunogen
          Hemopexin purified from human plasma.
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1.11 mg/mL
          Amount
          166.5 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.4, with 0.5M NaCl
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_667475
          Product Specific Information
          ABS 013-04B-01 detects Hemopexin in human samples.
          ABS 013-04B-01 has been successfully used in ELISA procedures.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
          Target Information
          Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.
          Bioinformatics
          Protein Aliases: Beta-1B-glycoprotein; epididymis secretory sperm binding protein; Hemopexin; hemopexin precursor; unnamed protein product
          View more View less
          Gene Aliases: HPX; HX
          View more View less
          UniProt ID: (Human) P02790
          View more View less
          Entrez Gene ID: (Human) 3263
          View more View less
          Function(s)
          protein binding heme transporter activity metal ion binding
          Process(es)
          positive regulation of immunoglobulin production positive regulation of humoral immune response mediated by circulating immunoglobulin cellular iron ion homeostasis heme transport hemoglobin metabolic process heme metabolic process regulation of protein metabolic process positive regulation of response to interferon-gamma interferon-gamma-mediated signaling pathway positive regulation of interferon-gamma-mediated signaling pathway
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline